Search
Search

Dhx30| Gene Symbol: Dhx30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252682.1 | NP_001239611.1 | 6 - 7 | 823 | 70 | |
| NM_001252683.1 | NP_001239612.1 | 2 - 3 | 402 | 70 | ||
| NM_133347.2 | NP_579925.1 | 5 - 6 | 499 | 70 | ||
| GenBank mRNA | AB047557.1 | - | 5 - 6 | 499 | 70 | |
| AK048409.1 | - | 3 - 4 | 1517 | 70 | ||
| AK122395.1 | - | 3 - 4 | 467 | 70 | ||
| AK146326.1 | - | 6 - 7 | 630 | 70 | ||
| AK153969.1 | - | 6 - 7 | 586 | 70 | ||
| AK159601.1 | - | 6 - 7 | 621 | 70 | ||
| AK164425.1 | - | 2 - 3 | 1100 | 70 | ||
| AK191102.1 | - | 1 - 2 | 103 | 70 | ||
| AK201136.1 | - | 1 - 2 | 103 | 70 | ||
| AK215800.1 | - | 1 - 2 | 103 | 70 | ||
| BC016202.1 | - | 2 - 3 | 348 | 4527765 | 70 | |
| BC058331.1 | - | 4 - 5 | 902 | 70 | ||
| BC062885.1 | - | 6 - 7 | 1190 | 70 | ||
| BC084729.1 | - | 4 - 5 | 463 | 70 | ||
Set Membership: |
|