Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00504796_m1
          See other OOSP1 GE Assays ›
          Gene Symbol:
          Oosp1
          Gene Name:
          oocyte secreted protein 1
          Gene Aliases:
          AA536827
          Chromosome Location:
          Chr.19: 11667460 - 11691051 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00504796_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Oosp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133353.3 NP_579931.1 1 - 2 106 76
          GenBank mRNA AB050982.2 - 1 - 2 99 76
          AB086437.1 - 1 - 2 89 76
          AF420487.1 - 1 - 2 111 76
          AK033038.1 - 1 - 2 109 76
          AK162132.1 - 1 - 2 109 76
          BC100313.1 - 1 - 2 85 30936071 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900001142.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Oosp1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P195135 AK033038.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P195135 AK162132.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P195135 BC100313.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P195135 NM_133353.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P195135 AB050982.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P195135 AK033038.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P195135 AK162132.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P195135 BC100313.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P195135 NM_133353.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P195135 AB050982.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2013431 AK033038.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2013431 AK162132.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2013431 BC100313.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2013431 NM_133353.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2013431 AB050982.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 140542 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 109684 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 859297 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 234061 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 29176 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 69135 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P179877 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P111491 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2013431 AK033038.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2013431 AK162132.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2013431 BC100313.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2013431 NM_133353.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2013431 AB050982.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P195135 AK033038.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P195135 AK162132.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P195135 BC100313.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P195135 NM_133353.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P195135 AB050982.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P195135 AK033038.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P195135 AK162132.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P195135 BC100313.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P195135 NM_133353.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P195135 AB050982.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31957282 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31957309 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31957288 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31957297 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31957305 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31957314 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31957321 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31957285 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31957298 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31957292 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31957289 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31957311 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31957306 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31957308 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31957287 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31957304 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31957319 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31957301 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31957303 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31957300 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31957293 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31957315 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03550889 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03550890 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P179877 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P111491 - -
          Whole Mouse Genome G4122A A_52_P195135 AK033038.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P195135 AK162132.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P195135 BC100313.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P195135 NM_133353.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P195135 AB050982.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P195135 AK033038.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P195135 AK162132.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P195135 BC100313.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P195135 NM_133353.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P195135 AB050982.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P205042 AK033038.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P205042 AK162132.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P205042 BC100313.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P205042 NM_133353.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P205042 AB050982.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134435 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117985 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P111491 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P179877 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14485629 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14485635 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14277584 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14485632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31957249 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14189684 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31957272 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31957287 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31957274 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31957285 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31957301 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31957293 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31957297 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31957268 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31957292 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31957288 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31957281 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31957311 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14302952 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31957300 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31957276 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31957275 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31957266 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31957247 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31957304 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14302957 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31957309 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31957291 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31957305 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31957303 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31957264 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31957280 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31957289 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31957306 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31957261 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31957260 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31957270 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31957298 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31957308 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31957249 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10466244 - -
          Affymetrix Mouse Gene 1.0 ST Array 10466242 - -
          Affymetrix Mouse Gene 1.0 ST Array 10466246 - -
          Affymetrix Mouse Gene 1.0 ST Array 10466245 - -
          Affymetrix Mouse Gene 1.0 ST Array 10466243 - -
          Affymetrix Mouse Gene 1.0 ST Array 10466241 - -
          Affymetrix Mouse Gene 1.0 ST Array 10466247 - -
          Affymetrix Mouse Gene 1.0 ST Array 10466240 - -
          Affymetrix HT MG-430 PM Array Plate 1418531_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10466239 - -
          Mouse Genome 430 2.0 Array 1418531_PM_at - -
          Mouse Genome 430 2.0 Array 1418531_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 170834_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 170834_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1900010724.mm.1 NM_133353.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010724.mm.1 NM_133353.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010724.mm.1 TC1900001142.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900010723.mm.1 NM_133353.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010723.mm.1 NM_133353.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010723.mm.1 TC1900001142.mm.1 MTA TC
          Mouse Expression Set 430 1418531_at - -
          Mouse Expression Set 430 1418531_PM_at - -
          Mouse Genome 430A 2.0 Array 1418531_at - -
          Mouse Genome 430A 2.0 Array 1418531_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418531_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418531_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 428949 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3440008 - -
          Illumina mouse-6 v1.0 expression beadchip 3440008 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003440008 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2707996 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2707996 - -
          mouse-6 v1.1 (Illumina) scl52718.5.1_51-S - -
          Illumina Sentrix mouse-6.v1 bead array 3440008 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl52718.5.1_51-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3440008 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2707996 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2707996 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline