Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00504831_m1
          See other ZFP617 GE Assays ›
          Gene Symbol:
          Zfp617
          Gene Name:
          zinc finger protein 617
          Gene Aliases:
          Zfps11-6
          Chromosome Location:
          Chr.8: 71922825 - 71934630 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00504831_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp617
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133358.3 NP_579936.2 3 - 4 333 131
          GenBank mRNA AB020542.1 - 294 131
          AK143880.1 - 3 - 4 307 131
          BC014712.1 - 3 - 4 239 4507398 131

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800003231.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp617

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25959078 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20409778 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20409779 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20409776 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20409781 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20409777 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25959075 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25959077 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25959079 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2043668 AK146644.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2043668 BX519071.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2043668 AK143880.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2043668 NM_133358.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 511734 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 186918 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 44347 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 575575 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 429482 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 141403 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P160810 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P174095 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2043668 AK146644.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2043668 BX519071.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2043668 AK143880.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2043668 NM_133358.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959073 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959054 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959058 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959081 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959075 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959068 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959050 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959071 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959079 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959055 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959065 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959052 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959064 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959056 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959057 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959088 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959084 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959051 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959070 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959062 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959085 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959086 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959082 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25959060 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P174095 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06552134 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P160810 - -
          Whole Mouse Genome G4122A A_52_P586090 AK146644.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P586090 BC014712.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P586090 AB020542.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P586090 AK143880.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P586090 NM_133358.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P188189 AK146644.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P188189 BC014712.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P188189 AB020542.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P188189 AK143880.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P188189 NM_133358.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118211 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P160810 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P174095 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06829535 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06829536 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06829531 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06829533 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25959077 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20409779 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20409781 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20409777 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20409776 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25959079 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25959078 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20409778 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25959075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25959081 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06829535 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959047 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959038 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959042 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959083 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959058 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959025 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959035 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959098 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959040 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959051 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959081 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06976489 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959054 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959093 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06696500 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06829537 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959055 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959057 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959052 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07314768 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959099 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959048 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959100 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959095 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959091 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959028 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959102 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959088 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959082 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06737869 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959071 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959060 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959105 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959066 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959046 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07412991 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959106 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25959085 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10572731 - -
          Affymetrix Mouse Gene 1.0 ST Array 10572732 - -
          Affymetrix HT MG-430 PM Array Plate 1449546_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1434355_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10572730 - -
          Mouse Genome 430 2.0 Array 1434355_PM_at - -
          Mouse Genome 430 2.0 Array 1449546_a_at - -
          Mouse Genome 430 2.0 Array 1443640_at - -
          Mouse Genome 430 2.0 Array 1443931_PM_at - -
          Mouse Genome 430 2.0 Array 1443640_PM_at - -
          Mouse Genome 430 2.0 Array 1449546_PM_a_at - -
          Mouse Genome 430 2.0 Array 1434355_at - -
          Mouse Genome 430 2.0 Array 1443931_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 170938_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 170938_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0800024767.mm.1 NM_133358.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800024767.mm.1 NM_133358.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800024767.mm.1 TC0800003231.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800005913.mm.1 NM_133358.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005913.mm.1 NM_133358.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005913.mm.1 TC0800003231.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800005917.mm.1 NM_133358.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005917.mm.1 NM_133358.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005917.mm.1 TC0800003231.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800005911.mm.1 NM_133358.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005911.mm.1 NM_133358.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005911.mm.1 TC0800003231.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800005920.mm.1 NM_133358.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005920.mm.1 NM_133358.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005920.mm.1 TC0800003231.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800005914.mm.1 NM_133358.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005914.mm.1 NM_133358.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005914.mm.1 TC0800003231.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800005916.mm.1 NM_133358.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005916.mm.1 NM_133358.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005916.mm.1 TC0800003231.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800005912.mm.1 NM_133358.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005912.mm.1 NM_133358.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005912.mm.1 TC0800003231.mm.1 MTA TC
          Mouse Expression Set 430 1449546_a_at - -
          Mouse Expression Set 430 1449546_PM_a_at - -
          Mouse Expression Set 430 1443640_PM_at - -
          Mouse Expression Set 430 1443640_at - -
          Mouse Expression Set 430 1434355_PM_at - -
          Mouse Expression Set 430 1434355_at - -
          Murine Genome U74v2 Set 100713_at - -
          Murine Genome U74v2 Set 100713_PM_at - -
          Mouse Genome 430A 2.0 Array 1449546_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1449546_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1449546_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1449546_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1434355_at - -
          Affymetrix HT Mouse Genome 430B Array 1443640_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1434355_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1443640_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2030315 - -
          Illumina mouse-6 v1.0 expression beadchip 2030315 - -
          Illumina mouse-6 v1.0 expression beadchip 3290286 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002030315 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003290286 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1236104 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2468999 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1236104 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2468999 - -
          mouse-6 v1.1 (Illumina) scl00170938.1_81-S - -
          mouse-6 v1.1 (Illumina) scl0170938.6_163-S - -
          Illumina Sentrix mouse-6.v1 bead array 3290286 - -
          Illumina Sentrix mouse-6.v1 bead array 2030315 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00170938.1_81-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0170938.6_163-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3290286 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2030315 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2503418 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2503420 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2503418 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2503420 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline