| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23318023 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23318022 |
- |
- |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1971991 |
AK158383.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1971991 |
BC006890.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1971991 |
BF318332.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1971991 |
BG069889.2 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1971991 |
NM_001083916.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
249367 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
742302 |
AK035231.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
742302 |
AK078773.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
742302 |
AK154569.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
742302 |
AK155150.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
742302 |
AK155752.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
742302 |
AK157265.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
742302 |
BC052163.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
742302 |
NM_007961.3 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
742302 |
Y07915.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
932304 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
554520 |
AK034730.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
792430 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
901994 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
618351 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
48344 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P162620 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P111489 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1971991 |
AK158383.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1971991 |
BC006890.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1971991 |
BF318332.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1971991 |
BG069889.2 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1971991 |
NM_001083916.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23318017 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23318008 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23318024 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23318028 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23318005 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23318030 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23318022 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23318003 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23318021 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23318013 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23318020 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23318012 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23318007 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23318025 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23318023 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23318009 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23318010 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23318015 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23318019 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23318027 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23318002 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P162620 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P111489 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05409588 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05409580 |
- |
- |
| Whole Mouse Genome G4122A |
A_51_P142258 |
AK158383.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P142258 |
BC006890.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P142258 |
BF318332.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P142258 |
BG069889.2 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P142258 |
NM_001083916.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P124450 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P111489 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P162620 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03858260 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03409218 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03858256 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23318023 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23318022 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318012 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318089 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23317992 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318091 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23317997 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318016 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318114 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318067 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318119 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318122 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318030 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318008 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318066 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318126 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318061 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318032 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318063 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318054 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318137 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318077 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318017 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318027 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318021 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318059 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318014 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23317996 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318079 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318035 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318070 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23317994 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318090 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318104 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318007 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318046 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318096 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318121 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318020 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318025 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318039 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318127 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318125 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318049 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23317999 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318112 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318106 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318110 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318019 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318031 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318057 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318116 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318123 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318134 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318095 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318022 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318050 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23317991 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318129 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318062 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318135 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318042 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318074 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318023 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318075 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318002 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318072 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318093 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318069 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23318024 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03759610 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23318006 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23318045 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23318010 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03759607 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23318002 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23318016 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03219509 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23317996 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03764153 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23318032 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03968559 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03971727 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04091021 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23318040 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23318008 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03968560 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23318031 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23318017 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03568807 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23318042 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23318039 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03920514 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03759611 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23318019 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03932486 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23317992 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23318035 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03566834 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23318041 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23317994 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23318005 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03530533 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23318012 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03409218 |
- |
- |