Search
Search

Ctdspl| Gene Symbol: Ctdspl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133710.3 | NP_598471.3 | 2 - 3 | 561 | 69 | |
| GenBank mRNA | AA239542.1 | - | 2 - 3 | 252 | 69 | |
| AJ344340.1 | - | 2 - 3 | 643 | 69 | ||
| AK013115.1 | - | 2 - 3 | 557 | 69 | ||
| BC094289.1 | - | 2 - 3 | 246 | 4986338 | 69 | |
Set Membership: |
|