Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00505209_m1
          See other PRSS22 GE Assays ›
          Gene Symbol:
          Prss22
          Gene Name:
          protease, serine 22
          Gene Aliases:
          4733401N09Rik, BSSP-4, SP001LA
          Chromosome Location:
          Chr.17: 23993534 - 24000735 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00505209_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prss22
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133731.2 NP_598492.2 4 - 5 654 90
          GenBank mRNA AB010778.1 - 4 - 5 637 90
          AK014645.1 - 4 - 5 654 90
          BC055854.1 - 4 - 5 639 4035351 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700001612.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prss22

          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          serine-type endopeptidase activity
          peptidase activity
          serine-type peptidase activity
          hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P121590 NM_133731.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P121590 AB010778.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P121590 AK014645.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P121590 BC055854.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 183011 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 938903 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P175202 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P121590 NM_133731.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P121590 AB010778.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P121590 AK014645.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P121590 BC055854.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P363258 CD773447.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P363258 NM_133731.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P363258 AB010778.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P363258 BC055854.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094784 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094799 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094785 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094789 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094787 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094815 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094802 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094795 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094800 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094798 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094825 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094821 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094796 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094816 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094819 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094813 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094804 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094824 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094806 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094809 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094794 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094791 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094823 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094817 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094807 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094803 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094792 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094786 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094801 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094790 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094811 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P175202 - -
          Whole Mouse Genome G4122A A_51_P363258 CD773447.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P363258 NM_133731.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P363258 AB010778.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P363258 BC055854.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121590 NM_133731.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121590 AB010778.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121590 AK014645.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121590 BC055854.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P175202 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13555971 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13555969 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13555979 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13555970 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13555966 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13555976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094845 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094743 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094764 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094786 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13063831 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094815 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094762 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094789 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094745 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094803 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094813 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094796 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094811 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094825 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094778 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094818 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094838 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13074373 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094806 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094819 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13462644 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094823 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094794 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094845 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094748 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094759 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094826 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13462645 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13215497 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094804 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094754 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094773 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094757 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094800 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094770 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13511512 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094840 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094809 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094801 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13462652 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13296551 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13276195 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094767 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13511511 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13063830 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13074374 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094842 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094785 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094816 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094753 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094807 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094797 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094827 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13074376 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094828 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094817 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13462650 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13282106 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094798 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094741 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13511513 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13346807 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094752 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10448410 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448411 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448413 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448414 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448412 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448415 - -
          Affymetrix HT MG-430 PM Array Plate 1420352_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10448409 - -
          Mouse Genome 430 2.0 Array 1420352_at - -
          Mouse Genome 430 2.0 Array 1420352_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70835_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70835_at - -
          Affymetrix Mouse Clariom S Array PSR1700014144.mm.1 NM_133731.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014144.mm.1 NM_133731.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014144.mm.1 TC1700001612.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700014139.mm.1 NM_133731.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014139.mm.1 NM_133731.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014139.mm.1 TC1700001612.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700014140.mm.1 NM_133731.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014140.mm.1 NM_133731.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014140.mm.1 TC1700001612.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700014142.mm.1 NM_133731.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014142.mm.1 NM_133731.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014142.mm.1 TC1700001612.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700014141.mm.1 NM_133731.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014141.mm.1 NM_133731.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014141.mm.1 TC1700001612.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700014143.mm.1 NM_133731.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014143.mm.1 NM_133731.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014143.mm.1 TC1700001612.mm.1 MTA TC
          Mouse Expression Set 430 1420352_at - -
          Mouse Expression Set 430 1420352_PM_at - -
          Mouse Genome 430A 2.0 Array 1420352_at - -
          Mouse Genome 430A 2.0 Array 1420352_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1420352_at - -
          Affymetrix HT Mouse Genome 430A Array 1420352_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 578302 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4210368 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004210368 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2688287 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2688287 - -
          mouse-6 v1.1 (Illumina) scl070835.2_8-S - -
          Illumina Sentrix mouse-6.v1 bead array 4210368 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl070835.2_8-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4210368 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2688287 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2688287 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline