Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00505236_m1
          See other PTCD1 GE Assays ›
          Gene Symbol:
          Ptcd1
          Gene Name:
          pentatricopeptide repeat domain 1
          Gene Aliases:
          1110069M14Rik
          Chromosome Location:
          Chr.5: 145147378 - 145167119 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00505236_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ptcd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133735.2 NP_598496.3 2 - 3 630 69
          GenBank mRNA AK088764.1 - 2 - 3 549 69
          AK143877.1 - 2 - 3 587 69
          AK150514.1 - 2 - 3 587 69
          AK151064.1 - 2 - 3 599 69
          AK152077.1 - 2 - 3 599 69
          AK153366.1 - 2 - 3 599 69
          AK171484.1 - 2 - 3 585 69
          BC004766.1 - 2 - 3 548 3156304 69
          BC083097.1 - 2 - 3 589 30459747 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500003590.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ptcd1

          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          tRNA processing
          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24164619 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24164621 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24164620 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2021545 AK172986.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P163079 BC083097.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P163079 BC004766.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P163079 AK171484.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P163079 AK166306.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P163079 AK152077.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P163079 BC083097.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P163079 BC004766.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P163079 AK171484.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P163079 AK166306.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P163079 AK152077.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 77942 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 438612 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 568111 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 634639 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 393792 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 946541 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 132248 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 267441 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 201198 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 815645 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 317432 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 169620 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 82092 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P154410 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P126342 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2021545 AK172986.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P163079 BC083097.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P163079 BC004766.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P163079 AK171484.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P163079 AK166306.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P163079 AK152077.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P163079 BC083097.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P163079 BC004766.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P163079 AK171484.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P163079 AK166306.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P163079 AK152077.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24164602 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24164613 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24164612 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24164614 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24164609 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24164617 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24164608 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24164619 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24164620 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24164611 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24164610 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01243926 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01243938 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P126342 - -
          Whole Mouse Genome G4122A A_52_P227065 BC006861.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P227065 BC006861.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P163079 BC083097.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P163079 BC004766.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P163079 AK171484.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P163079 AK166306.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P163079 AK152077.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P163079 BC083097.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P163079 BC004766.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P163079 AK171484.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P163079 AK166306.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P163079 AK152077.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133320 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P126342 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04923171 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04139410 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04923170 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04923169 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24164621 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24164619 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24164620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24164505 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04591839 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24164614 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24164588 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24164617 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24164600 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24164592 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04923169 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24164596 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24164593 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24164598 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04680664 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24164595 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24164609 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24164590 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24164615 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24164594 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24164597 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04200223 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24164606 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04985267 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24164610 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04680660 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24164619 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04923170 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10527454 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535643 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535641 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535640 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535646 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535642 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535644 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535645 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535639 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535638 - -
          Affymetrix HT MG-430 PM Array Plate 1425096_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1416391_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10527448 - -
          Affymetrix Mouse Gene 1.1 ST Array 10535637 - -
          Mouse Genome 430 2.0 Array 1425096_a_at - -
          Mouse Genome 430 2.0 Array 1416391_at - -
          Mouse Genome 430 2.0 Array 1425096_PM_a_at - -
          Mouse Genome 430 2.0 Array 1416391_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71799_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71799_at - -
          Affymetrix Mouse Clariom S Array PSR0500032936.mm.1 TC0500003590.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500032936.mm.1 NM_133735.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500032936.mm.1 NM_133735.2 Refseq
          Mouse Expression Set 430 1425096_PM_a_at - -
          Mouse Expression Set 430 1425096_a_at - -
          Mouse Expression Set 430 1416391_at - -
          Mouse Expression Set 430 1416391_PM_at - -
          Mouse Genome 430A 2.0 Array 1425096_a_at - -
          Mouse Genome 430A 2.0 Array 1425096_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1416391_at - -
          Mouse Genome 430A 2.0 Array 1416391_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1425096_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1416391_at - -
          Affymetrix HT Mouse Genome 430A Array 1416391_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1425096_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 904724 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-vppth:80/100.66.77.159:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline