Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00505601_m1
          See other NUSAP1 GE Assays ›
          Gene Symbol:
          Nusap1
          Gene Name:
          nucleolar and spindle associated protein 1
          Gene Aliases:
          2610201A12Rik, AI481307, ANKT, AW547774, BB165529, BM037, LNP, NuSAP, Q0310, SAPL
          Chromosome Location:
          Chr.2: 119618298 - 119650160 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00505601_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nusap1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001042652.1 NP_001036117.1 1 - 2 600 58
          NM_133851.3 NP_598612.1 1 - 2 600 58
          GenBank mRNA AF305710.1 - 1 - 2 181 58
          AK042697.1 - 181 58
          AK044819.1 - 1 - 2 171 58
          AK076028.1 - 1 - 2 170 58
          AK088389.1 - 1 - 2 170 58
          BC004787.1 - 1 - 2 150 3588697 58
          BC100392.1 - 1 - 2 600 30110191 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200001776.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nusap1

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic sister chromatid segregation
          microtubule cytoskeleton organization
          mitotic cytokinesis
          cell cycle
          mitotic nuclear division
          mitotic chromosome condensation
          establishment of mitotic spindle localization
          positive regulation of mitotic nuclear division
          cell division
          DNA binding
          microtubule binding
          poly(A) RNA binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200013339.mm.1 NM_001042652.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013339.mm.1 NM_001042652.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013339.mm.1 NM_133851.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013339.mm.1 NM_133851.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013339.mm.1 TC0200001776.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200013329.mm.1 NM_001042652.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013329.mm.1 NM_001042652.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013329.mm.1 NM_133851.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013329.mm.1 NM_133851.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013329.mm.1 TC0200001776.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200013334.mm.1 NM_001042652.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013334.mm.1 NM_001042652.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013334.mm.1 NM_133851.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013334.mm.1 NM_133851.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013334.mm.1 TC0200001776.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 932873 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline