Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00505856_m1
          See other LSM8 GE Assays ›
          Gene Symbol:
          Lsm8
          Gene Name:
          LSM8 homolog, U6 small nuclear RNA associated (S. cerevisiae)
          Gene Aliases:
          2010003I05Rik, AW214405, Naa38
          Chromosome Location:
          Chr.6: 18848635 - 18854052 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00505856_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lsm8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133939.1 NP_598700.1 3 - 4 292 92
          GenBank mRNA AK088664.1 - 3 - 4 253 92
          AK154446.1 - 1157 92
          AK167674.1 - 224 92
          BC019458.1 - 3 - 4 292 4024697 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600000151.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lsm8

          Panther Classification:

          Molecular Function -

          RNA splicing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA splicing, via spliceosome
          mRNA processing
          biological_process
          RNA splicing
          RNA binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24275503 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24275505 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24275502 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P306658 AK154446.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P306658 AK167674.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P306658 BC019458.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P306658 NM_133939.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P306658 AK088664.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P306658 AK154446.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P306658 AK167674.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P306658 BC019458.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P306658 NM_133939.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P306658 AK088664.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 26117 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 572819 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 424042 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 648286 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 102923 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P180051 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P306658 AK154446.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P306658 AK167674.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P306658 BC019458.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P306658 NM_133939.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P306658 AK088664.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P306658 AK154446.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P306658 AK167674.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P306658 BC019458.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P306658 NM_133939.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P306658 AK088664.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P306658 AK154446.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P306658 AK167674.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P306658 BC019458.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P306658 NM_133939.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P306658 AK088664.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P306658 AK154446.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P306658 AK167674.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P306658 BC019458.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P306658 NM_133939.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P306658 AK088664.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24275502 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24275510 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24275500 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24275505 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24275508 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24275511 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24275519 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24275520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24275515 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24275503 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24275516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24275521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24275513 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24275507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24275523 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05825899 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P180051 - -
          Whole Mouse Genome G4122A A_51_P306658 AK154446.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P306658 AK167674.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P306658 BC019458.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P306658 NM_133939.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P306658 AK088664.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P306658 AK154446.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P306658 AK167674.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P306658 BC019458.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P306658 NM_133939.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P306658 AK088664.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115976 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P180051 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05184640 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05184642 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24275502 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24275505 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24275503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275476 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275511 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275515 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05296104 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275532 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275547 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05476593 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275521 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05296105 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275489 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275500 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05476604 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275519 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05599521 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275508 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275484 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275549 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05097387 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275539 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275529 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275559 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275510 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05389437 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05097385 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275523 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275551 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275552 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275554 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275543 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275556 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05476601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275487 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275502 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275505 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275492 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05296115 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275524 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275520 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275536 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275499 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275486 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275516 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275525 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275541 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10536599 - -
          Affymetrix Mouse Gene 1.0 ST Array 10536597 - -
          Affymetrix Mouse Gene 1.0 ST Array 10536598 - -
          Affymetrix Mouse Gene 1.0 ST Array 10536596 - -
          Affymetrix HT MG-430 PM Array Plate 1436107_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1448703_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10536595 - -
          Mouse Genome 430 2.0 Array 1448703_PM_at - -
          Mouse Genome 430 2.0 Array 1448703_at - -
          Mouse Genome 430 2.0 Array 1436107_PM_at - -
          Mouse Genome 430 2.0 Array 1436107_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76522_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76522_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600001068.mm.1 NM_133939.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600001068.mm.1 NM_133939.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600001068.mm.1 TC0600000151.mm.1 MTA TC
          Mouse Expression Set 430 1448703_at - -
          Mouse Expression Set 430 1448703_PM_at - -
          Mouse Expression Set 430 1436107_at - -
          Mouse Expression Set 430 1436107_PM_at - -
          Murine Genome U74v2 Set 102409_PM_at - -
          Murine Genome U74v2 Set 102409_at - -
          Mouse Genome 430A 2.0 Array 1448703_at - -
          Mouse Genome 430A 2.0 Array 1448703_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448703_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448703_at - -
          Affymetrix HT Mouse Genome 430B Array 1436107_at - -
          Affymetrix HT Mouse Genome 430B Array 1436107_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 631716 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 460068 - -
          Illumina mouse-6 v1.0 expression beadchip 460068 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000460068 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1230299 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1230299 - -
          mouse-6 v1.1 (Illumina) scl30348.4.1_28-S - -
          Illumina Sentrix mouse-6.v1 bead array 460068 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl30348.4.1_28-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 460068 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2883907 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2883907 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl30348.4.1_28-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline