Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00506001_m1
          See other SNAPC2 GE Assays ›
          Gene Symbol:
          Snapc2
          Gene Name:
          small nuclear RNA activating complex, polypeptide 2
          Gene Aliases:
          0610007H10Rik, AU015675
          Chromosome Location:
          Chr.8: 4253102 - 4256220 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00506001_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Snapc2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133968.1 NP_598729.1 1 - 2 201 125
          GenBank mRNA AK035340.1 - 1 - 2 213 125
          AK075569.1 - 1 - 2 224 125
          AK141475.1 - 1 - 2 203 125
          AK146218.1 - 1 - 2 193 125
          AK155012.1 - 1 - 2 213 125
          BC011147.1 - 1 - 2 201 4194905 125

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800000032.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Snapc2

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25616848 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25616821 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25616824 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25616846 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25616822 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25616820 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P109315 BC011147.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P109315 AK146218.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P109315 AK155012.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P109315 AK075569.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P109315 AK035340.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 332998 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P161337 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P109315 BC011147.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P109315 AK146218.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P109315 AK155012.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P109315 AK075569.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P109315 AK035340.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25616811 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25616860 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25616809 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25616810 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25616823 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25616832 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25616836 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25616829 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25616854 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25616847 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25616848 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25616842 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25616852 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25616849 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25616824 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25616828 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25616815 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25616865 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25616826 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25616840 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25616835 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25616844 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25616808 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25616846 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25616850 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25616817 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25616859 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25616856 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25616830 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25616876 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25616861 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25616838 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25616822 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25616816 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25616818 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25616839 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25616820 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25616813 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P161337 - -
          Whole Mouse Genome G4122A A_51_P269153 BC011147.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P269153 AK186997.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P269153 AK146218.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P269153 AK155012.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P269153 AK181477.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138735 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109315 BC011147.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109315 AK146218.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109315 AK155012.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109315 AK075569.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109315 AK035340.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P161337 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07124553 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07123477 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07203089 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25616821 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25616822 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25616846 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25616820 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25616824 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25616848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616822 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616876 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616865 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616823 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07123477 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616845 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616840 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616835 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616866 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616853 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616836 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616844 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06771790 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616828 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616861 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616850 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616810 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616842 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616826 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616815 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616817 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07200063 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616870 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06771755 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06771753 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07200069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616854 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616871 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616813 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616856 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616816 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616875 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616822 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06771746 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616873 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616820 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06771787 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10569955 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418298 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569954 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569957 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569956 - -
          Affymetrix HT MG-430 PM Array Plate 1436703_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1448726_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10418297 - -
          Affymetrix Mouse Gene 1.1 ST Array 10569953 - -
          Mouse Genome 430 2.0 Array 1436703_x_at - -
          Mouse Genome 430 2.0 Array 1436703_PM_x_at - -
          Mouse Genome 430 2.0 Array 1448726_PM_at - -
          Mouse Genome 430 2.0 Array 1448726_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 102209_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 102209_at - -
          Affymetrix Mouse Clariom S Array PSR0800000406.mm.1 NM_133968.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800000406.mm.1 NM_133968.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800000406.mm.1 TC0800000032.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800000407.mm.1 NM_133968.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800000407.mm.1 NM_133968.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800000407.mm.1 TC0800000032.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800000408.mm.1 NM_133968.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800000408.mm.1 NM_133968.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800000408.mm.1 TC0800000032.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800000410.mm.1 NM_133968.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800000410.mm.1 NM_133968.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800000410.mm.1 TC0800000032.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800000411.mm.1 NM_133968.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800000411.mm.1 NM_133968.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800000411.mm.1 TC0800000032.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800000409.mm.1 NM_133968.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800000409.mm.1 NM_133968.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800000409.mm.1 TC0800000032.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800000412.mm.1 NM_133968.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800000412.mm.1 NM_133968.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800000412.mm.1 TC0800000032.mm.1 MTA TC
          Mouse Expression Set 430 1436703_x_at - -
          Mouse Expression Set 430 1448726_at - -
          Mouse Expression Set 430 1436703_PM_x_at - -
          Mouse Expression Set 430 1448726_PM_at - -
          Murine Genome U74v2 Set 160294_PM_at - -
          Murine Genome U74v2 Set 160294_at - -
          Mouse Genome 430A 2.0 Array 1436703_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1436703_x_at - -
          Mouse Genome 430A 2.0 Array 1448726_PM_at - -
          Mouse Genome 430A 2.0 Array 1448726_at - -
          Affymetrix HT Mouse Genome 430A Array 1436703_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1448726_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1436703_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1448726_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 919637 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4060180 - -
          Illumina mouse-6 v1.0 expression beadchip 4060180 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004060180 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1215174 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1215174 - -
          mouse-6 v1.1 (Illumina) scl34118.6.1_7-S - -
          Illumina Sentrix mouse-6.v1 bead array 4060180 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl34118.6.1_7-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4060180 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1215174 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1215174 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl34118.6.1_7-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline