Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00506006_m1
          See other ANKRD10 GE Assays ›
          Gene Symbol:
          Ankrd10
          Gene Name:
          ankyrin repeat domain 10
          Gene Aliases:
          4833425P12Rik, AW549277
          Chromosome Location:
          Chr.8: 11611581 - 11636136 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00506006_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ankrd10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001167967.2 NP_001161439.1 2 - 3 503 74
          NM_001281974.1 NP_001268903.1 2 - 3 503 74
          NM_001281975.1 NP_001268904.1 2 - 3 503 74
          NM_133971.3 NP_598732.2 2 - 3 503 74
          NR_030781.2 2 - 3 382 74
          GenBank mRNA AK014768.1 - 2 - 3 382 74
          AK031808.1 - 2 - 3 501 74
          AK033623.1 - 2 - 3 501 74
          AK038156.1 - 2 - 3 501 74
          AK042500.1 - 2 - 3 501 74
          AK050983.1 - 2 - 3 504 74
          AK081754.1 - 2 - 3 501 74
          AK082875.1 - 2 - 3 500 74
          AK084456.1 - 2 - 3 501 74
          AK088371.1 - 2 - 3 453 74
          AK143253.1 - 2 - 3 501 74
          BC002198.1 - 2 - 3 435 3489133 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800001701.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the NM_001281974.1,NM_001281975.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ankrd10

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of canonical Wnt signaling pathway
          molecular_function

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0800013820.mm.1 NM_001281974.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013820.mm.1 NR_030781.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013820.mm.1 TC0800001701.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800013823.mm.1 NM_001167967.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013823.mm.1 NM_001281974.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013823.mm.1 NM_001281975.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013823.mm.1 NM_133971.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013823.mm.1 NR_030781.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013823.mm.1 TC0800001701.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline