Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00506297_s1
          See other AW209491 GE Assays ›
          Gene Symbol:
          AW209491
          Gene Name:
          expressed sequence AW209491
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 14630245 - 14638202 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00506297_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: AW209491
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001104646.1 NP_001098116.1 2 687 80
          NM_134067.4 NP_598828.3 2 754 80
          GenBank mRNA AK030506.1 - 736 80
          AK050507.1 - 2 736 80
          AK077649.1 - 2 736 80
          AK167401.1 - 2 755 80
          AK169400.1 - 2 687 80
          BC016098.1 - 1 651 4910571 80
          BC034309.1 - 1 510 5362794 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300000102.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene AW209491

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P400677 BC016098.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P400677 AK169400.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P400677 BC034309.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P400677 AK077649.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P400677 AK167401.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P400677 BC016098.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P400677 AK169400.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P400677 BC034309.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P400677 AK077649.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P400677 AK167401.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 885432 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 511448 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 969245 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 122103 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 135705 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P125470 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P400677 BC016098.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P400677 AK169400.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P400677 BC034309.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P400677 AK077649.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P400677 AK167401.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P400677 BC016098.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P400677 AK169400.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P400677 BC034309.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P400677 AK077649.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P400677 AK167401.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28839471 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28839489 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28839491 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28839482 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28839495 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28839475 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28839484 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28839505 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28839504 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28839486 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28839483 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28839498 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28839497 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28839503 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28839507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28839473 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28839476 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28839478 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28839501 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28839485 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28839470 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P125470 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07792446 - -
          Whole Mouse Genome G4122A A_52_P400677 BC016098.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P400677 AK169400.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P400677 BC034309.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P400677 AK077649.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P400677 AK167401.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P400677 BC016098.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P400677 AK169400.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P400677 BC034309.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P400677 AK077649.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P400677 AK167401.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127514 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P125470 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11055419 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10778236 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11055422 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10778231 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10528523 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11055420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839473 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839523 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839519 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839524 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839518 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839495 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839498 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839491 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839508 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839476 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839475 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839470 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839512 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839484 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839514 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839520 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11055414 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839479 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10537362 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839507 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10726319 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839473 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839528 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10523800 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839489 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839513 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839478 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10743459 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839516 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11055422 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839501 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10991850 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839485 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839527 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839517 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839521 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839471 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10778233 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10886930 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839480 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10886922 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10924975 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839511 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10403720 - -
          Affymetrix Mouse Gene 1.0 ST Array 10403719 - -
          Affymetrix Mouse Gene 1.0 ST Array 10403718 - -
          Affymetrix Mouse Gene 1.0 ST Array 10403717 - -
          Affymetrix HT MG-430 PM Array Plate 1423512_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10403716 - -
          Mouse Genome 430 2.0 Array 1423512_at - -
          Mouse Genome 430 2.0 Array 1423512_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 105351_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 105351_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1300000771.mm.1 NM_001104646.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000771.mm.1 NM_001104646.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000771.mm.1 NM_134067.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000771.mm.1 NM_134067.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000771.mm.1 TC1300000102.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300000768.mm.1 NM_001104646.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000768.mm.1 NM_001104646.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000768.mm.1 NM_134067.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000768.mm.1 NM_134067.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000768.mm.1 TC1300000102.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300000769.mm.1 NM_134067.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000769.mm.1 NM_134067.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000769.mm.1 TC1300000102.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300000773.mm.1 NM_001104646.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000773.mm.1 NM_001104646.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000773.mm.1 NM_134067.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000773.mm.1 NM_134067.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000773.mm.1 TC1300000102.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300000772.mm.1 NM_001104646.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000772.mm.1 NM_001104646.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000772.mm.1 NM_134067.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000772.mm.1 NM_134067.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000772.mm.1 TC1300000102.mm.1 MTA TC
          Mouse Expression Set 430 1423512_at - -
          Mouse Expression Set 430 1423512_PM_at - -
          Mouse Genome 430A 2.0 Array 1423512_at - -
          Mouse Genome 430A 2.0 Array 1423512_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423512_at - -
          Affymetrix HT Mouse Genome 430A Array 1423512_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 872246 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5290091 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005290091 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2717234 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2717234 - -
          mouse-6 v1.1 (Illumina) scl45053.3_267-S - -
          Illumina Sentrix mouse-6.v1 bead array 5290091 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl45053.3_267-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5290091 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2717234 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2717234 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline