Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00506520_m1
          See other NRM GE Assays ›
          Gene Symbol:
          Nrm
          Gene Name:
          nurim (nuclear envelope membrane protein)
          Gene Aliases:
          2610307M02Rik, AI429796
          Chromosome Location:
          Chr.17: 35861318 - 35865400 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00506520_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nrm
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_134122.2 NP_598883.1 1 - 2 202 86
          GenBank mRNA BC021750.1 - 1 - 2 205 4503599 86
          BC026576.1 - 1 - 2 174 4984796 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000706.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nrm

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31161344 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31161371 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31161378 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31161373 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31161345 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31161375 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31161380 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31161381 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31161382 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31161374 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31161346 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31161341 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31161377 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P379775 BC021750.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P379775 BC026576.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P379775 NM_134122.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P379775 AK143140.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P379775 BC021750.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P379775 BC026576.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P379775 NM_134122.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P379775 AK143140.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 850200 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 396656 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 278493 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P120564 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P379775 BC021750.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P379775 BC026576.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P379775 NM_134122.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P379775 AK143140.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P379775 BC021750.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P379775 BC026576.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P379775 NM_134122.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P379775 AK143140.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31161352 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31161325 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31161340 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31161320 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31161339 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31161318 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31161337 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31161348 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31161351 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31161335 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31161330 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31161331 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31161328 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31161333 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31161322 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31161327 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31161353 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31161341 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31161344 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31161324 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31161346 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31161350 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P120564 - -
          Whole Mouse Genome G4122A A_51_P379775 BC021750.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P379775 BC026576.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P379775 NM_134122.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P379775 AK143140.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P379775 BC021750.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P379775 BC026576.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P379775 NM_134122.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P379775 AK143140.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P313905 BC021750.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P313905 BC026576.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P313905 NM_134122.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P313905 AK143140.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138566 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P120564 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13320954 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13320956 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13256566 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13256568 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31161377 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31161381 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31161373 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31161378 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31161346 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31161345 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31161341 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31161382 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31161371 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31161344 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31161375 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31161374 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31161380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31161443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31161383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31161392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31161351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31161390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31161439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31161359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31161338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31161428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31161350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31161424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31161426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31161432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31161344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31161361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31161368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31161355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31161385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31161420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31161367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31161388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31161393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31161370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31161398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31161346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31161374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31161437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31161409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31161406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31161337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31161387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31161381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31161431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31161353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31161435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31161378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31161400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31161371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31161423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31161364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31161403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31161357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31161399 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31161398 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31161344 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31161355 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31161351 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31161383 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31161378 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31161400 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31161392 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31161350 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31161374 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31161388 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31161387 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13203131 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13256569 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13203130 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31161341 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31161368 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31161357 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31161367 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31161371 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31161380 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31161409 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31161393 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13437797 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13437790 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13523725 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31161370 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31161397 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31161348 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31161385 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13546249 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13320956 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31161406 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13203133 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13320954 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31161352 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31161336 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13437785 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31161359 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10444931 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444930 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444928 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444929 - -
          Affymetrix HT MG-430 PM Array Plate 1417971_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1443761_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10450611 - -
          Affymetrix Mouse Gene 1.1 ST Array 10444927 - -
          Mouse Genome 430 2.0 Array 1443761_PM_at - -
          Mouse Genome 430 2.0 Array 1417971_PM_at - -
          Mouse Genome 430 2.0 Array 1417971_at - -
          Mouse Genome 430 2.0 Array 1443761_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 106582_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 106582_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700007230.mm.1 NM_134122.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007230.mm.1 NM_134122.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007230.mm.1 TC1700000706.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700007231.mm.1 NM_134122.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007231.mm.1 NM_134122.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007231.mm.1 TC1700000706.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700007232.mm.1 NM_134122.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007232.mm.1 NM_134122.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007232.mm.1 TC1700000706.mm.1 MTA TC
          Mouse Expression Set 430 1417971_at - -
          Mouse Expression Set 430 1417971_PM_at - -
          Mouse Expression Set 430 1443761_at - -
          Mouse Expression Set 430 1443761_PM_at - -
          Murine Genome U74v2 Set 167249_PM_i_at - -
          Murine Genome U74v2 Set 167249_i_at - -
          Mouse Genome 430A 2.0 Array 1417971_at - -
          Mouse Genome 430A 2.0 Array 1417971_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417971_at - -
          Affymetrix HT Mouse Genome 430A Array 1417971_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1443761_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1443761_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 485013 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 520068 - -
          Illumina mouse-6 v1.0 expression beadchip 520068 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000520068 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2733524 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2733524 - -
          mouse-6 v1.1 (Illumina) scl50767.4_663-S - -
          Illumina Sentrix mouse-6.v1 bead array 520068 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl50767.4_663-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 520068 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2733524 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2733524 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl50767.4_663-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0