Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00507988_m1
          See other CD320 GE Assays ›
          Gene Symbol:
          Cd320
          Gene Name:
          CD320 antigen
          Gene Aliases:
          425O18-1, 8D6, D17Ertd716e, NG29, TCblR, VLDL
          Chromosome Location:
          Chr.17: 33843091 - 33849774 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00507988_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cd320
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019421.3 NP_062294.3 1 - 2 201 60
          GenBank mRNA AK021187.1 - 1 - 2 176 60
          AK078151.1 - 1 - 2 202 60
          AK088189.1 - 1 - 2 202 60
          AK140204.1 - 1 - 2 188 60
          BC026888.1 - 1 - 2 155 3991556 60
          KJ650205.1 - 1 - 2 121 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000595.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cd320

          Panther Classification:

          Molecular Function -

          apolipoprotein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of vitamin metabolic process
          growth factor activity
          cobalamin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31150731 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31150729 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31150732 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31150730 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2154425 AK140204.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2154425 NM_019421.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2154430 BC026888.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2154430 AK088189.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2154430 AK131655.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2154430 AK140204.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2154430 AK078151.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 873004 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 784484 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 644062 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 961596 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P173198 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2154425 AK140204.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2154425 NM_019421.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2154430 BC026888.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2154430 AK088189.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2154430 AK131655.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2154430 AK140204.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2154430 AK078151.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P337510 AK088189.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P337510 AK131655.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P337510 AK140204.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P337510 AK078151.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P337510 NM_019421.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P204744 BC026888.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P204744 AK088189.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P204744 AK131655.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P204744 AK140204.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P204744 AK078151.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31150745 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31150728 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31150719 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31150740 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31150736 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31150735 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31150733 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31150739 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31150721 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31150746 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31150744 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31150723 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31150747 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31150716 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31150724 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31150722 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31150727 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31150743 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31150718 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31150731 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P173198 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03309567 - -
          Whole Mouse Genome G4122A A_52_P204744 BC026888.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P204744 AK088189.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P204744 AK131655.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P204744 AK140204.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P204744 AK078151.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P337510 AK088189.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P337510 AK131655.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P337510 AK140204.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P337510 AK078151.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P337510 NM_019421.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115047 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P173198 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13578017 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13254176 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13254178 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13254177 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13254179 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13578010 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13578015 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31150729 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31150731 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31150732 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31150730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150749 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150770 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150718 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150722 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150753 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13446204 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150763 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13144740 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13092692 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150769 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150721 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150728 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150778 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150751 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150737 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150736 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150743 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150748 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13235991 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150739 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13471853 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150711 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150768 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150733 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13578017 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150776 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13254174 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150725 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150747 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150719 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150756 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150735 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150746 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150731 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13461894 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150749 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13471859 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13259074 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13471848 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150760 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150780 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150724 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150740 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10444051 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444055 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444050 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444054 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444047 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444048 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444053 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444052 - -
          Affymetrix HT MG-430 PM Array Plate 1417100_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1448588_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10444046 - -
          Mouse Genome 430 2.0 Array 1417100_at - -
          Mouse Genome 430 2.0 Array 1448588_PM_at - -
          Mouse Genome 430 2.0 Array 1448588_at - -
          Mouse Genome 430 2.0 Array 1417100_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 54219_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 54219_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700005476.mm.1 NM_019421.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005476.mm.1 NM_019421.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005476.mm.1 TC1700000595.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700005477.mm.1 NM_019421.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005477.mm.1 NM_019421.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005477.mm.1 TC1700000595.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700005479.mm.1 NM_019421.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005479.mm.1 NM_019421.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005479.mm.1 TC1700000595.mm.1 MTA TC
          Mouse Expression Set 430 1448588_at - -
          Mouse Expression Set 430 1417100_PM_at - -
          Mouse Expression Set 430 1417100_at - -
          Mouse Expression Set 430 1448588_PM_at - -
          Murine Genome U74v2 Set 102343_PM_at - -
          Murine Genome U74v2 Set 102343_at - -
          Mouse Genome 430A 2.0 Array 1448588_at - -
          Mouse Genome 430A 2.0 Array 1417100_at - -
          Mouse Genome 430A 2.0 Array 1417100_PM_at - -
          Mouse Genome 430A 2.0 Array 1448588_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448588_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417100_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417100_at - -
          Affymetrix HT Mouse Genome 430A Array 1448588_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 911668 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1450563 - -
          MouseWG-6 v2 Expression BeadChip 102190156 - -
          Illumina mouse-6 v1.0 expression beadchip 102190156 - -
          Illumina mouse-6 v1.0 expression beadchip 1450563 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001450563 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1235750 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1235750 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1237956 - -
          mouse-6 v1.1 (Illumina) scl0054219.2_27-S - -
          mouse-6 v1.1 (Illumina) scl50844.5_15-S - -
          Illumina Sentrix mouse-6.v1 bead array 102190156 - -
          Illumina Sentrix mouse-6.v1 bead array 1450563 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0054219.2_27-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102190156 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1450563 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1235750 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1235750 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline