Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00508266_m1
          See other 1700016D06RIK GE Assays ›
          Gene Symbol:
          1700016D06Rik
          Gene Name:
          RIKEN cDNA 1700016D06 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 11654921 - 11678750 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00508266_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700016D06Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024271.1 NP_077233.1 2 - 3 237 91
          GenBank mRNA AK006019.1 - 2 - 3 210 91
          AY026045.1 - 2 - 3 237 91
          BC048664.1 - 2 - 3 229 6773850 91

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800001703.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 1700016D06Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P306007 AY026045.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P306007 BC048664.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P306007 NM_024271.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P306007 AK006019.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P306007 AY026045.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P306007 BC048664.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P306007 NM_024271.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P306007 AK006019.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 483128 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 247310 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 896755 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 594177 BC048527.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 594177 NM_173367.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 345233 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 545976 NM_001009550.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 912209 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 310577 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 57258 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 285110 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 577628 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 523359 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 28615 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 902841 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 114008 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 96682 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 70332 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 108976 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P177026 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P134749 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P306007 AY026045.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P306007 BC048664.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P306007 NM_024271.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P306007 AK006019.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P306007 AY026045.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P306007 BC048664.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P306007 NM_024271.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P306007 AK006019.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25656873 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25656897 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25656884 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25656895 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25656881 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25656880 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25656863 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25656876 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25656870 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25656888 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25656869 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25656867 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25656862 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25656872 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25656878 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25656892 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25656886 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25656893 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25656877 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25656890 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25656885 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25656865 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25656866 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25656883 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06425208 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P134749 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06425161 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06425185 - -
          Whole Mouse Genome G4122A A_52_P306007 AY026045.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P306007 BC048664.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P306007 NM_024271.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P306007 AK006019.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P306007 AY026045.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P306007 BC048664.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P306007 NM_024271.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P306007 AK006019.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113489 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P134749 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06425161 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07002453 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07002472 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07002457 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07002478 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07002476 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07002455 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07002475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25656763 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25656881 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25656838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25656897 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25656867 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25656872 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25656841 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07043068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25656895 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25656876 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25656884 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07002453 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25656862 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25656907 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25656890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25656866 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25656910 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07002473 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25656906 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07021836 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25656883 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25656880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07021858 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25656836 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07214621 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25656861 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06762190 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25656887 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25656869 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25656865 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25656893 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25656839 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25656908 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25656913 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25656916 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07002457 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25656877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07002472 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25656889 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25656863 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10577058 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577062 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577059 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577060 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577061 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577063 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577064 - -
          Affymetrix HT MG-430 PM Array Plate 1425657_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10577057 - -
          Mouse Genome 430 2.0 Array 1425657_at - -
          Mouse Genome 430 2.0 Array 1425657_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76413_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76413_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0800013837.mm.1 NM_024271.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013837.mm.1 NM_024271.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013837.mm.1 TC0800001703.mm.1 MTA TC
          Mouse Expression Set 430 1425657_at - -
          Mouse Expression Set 430 1425657_PM_at - -
          Murine Genome U74v2 Set 131385_PM_at - -
          Murine Genome U74v2 Set 131385_at - -
          Mouse Genome 430A 2.0 Array 1425657_at - -
          Mouse Genome 430A 2.0 Array 1425657_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1425657_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1425657_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 584564 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1980497 - -
          Illumina mouse-6 v1.0 expression beadchip 1980497 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001980497 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2596795 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2596795 - -
          mouse-6 v1.1 (Illumina) scl35093.7.1_7-S - -
          Illumina Sentrix mouse-6.v1 bead array 1980497 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl35093.7.1_7-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1980497 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2939579 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2939583 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2939579 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2939583 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline