Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00508297_m1
          See other CCDC23 GE Assays ›
          Gene Symbol:
          Ccdc23
          Gene Name:
          coiled-coil domain containing 23
          Gene Aliases:
          2410005K17Rik, AI851162, Svbp
          Chromosome Location:
          Chr.4: 119195310 - 119201298 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00508297_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccdc23
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001038998.2 NP_001034087.1 2 - 3 396 89
          NM_024462.2 NP_077782.1 2 - 3 322 89
          GenBank mRNA AK010418.1 - 2 - 3 252 89
          AK079783.1 - 2 - 3 181 89
          BC002274.1 - 2 - 3 235 3496493 89
          BF012097.1 - 2 - 3 326 89
          BQ044357.1 - 1 - 2 88 89
          CF105092.1 - 2 - 3 382 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400001327.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ccdc23

          Gene Ontology Categories:

          Function(s) Process(es)

          protein secretion
          negative regulation of endothelial cell migration
          negative regulation of protein ubiquitination
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23244137 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23244138 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2155002 BQ044357.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2155002 NM_001038998.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2155002 NM_024462.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2155002 AK010418.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2155002 AK079783.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2169188 CF105092.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2169188 NM_001038998.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 346925 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 579679 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 239464 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 746939 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 36782 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P105705 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P104845 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P122426 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2155002 BQ044357.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2155002 NM_001038998.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2155002 NM_024462.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2155002 AK010418.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2155002 AK079783.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2169188 CF105092.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2169188 NM_001038998.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23244138 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23244152 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23244145 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23244153 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23244142 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23244148 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23244156 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23244150 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23244140 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23244147 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P122426 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P105705 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P104845 - -
          Whole Mouse Genome G4122A A_51_P303371 BQ044357.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P303371 NM_001038998.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P303371 NM_024462.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P303371 AK010418.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P303371 AK079783.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114182 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P105705 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P104845 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P122426 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23244137 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23244138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244102 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23244183 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23244130 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03347647 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23244150 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23244175 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03364939 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23244162 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03309658 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23244142 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23244173 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03820073 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23244186 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03816905 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03435341 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23244138 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23244168 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23244143 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23244147 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03347649 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23244129 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23244131 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03309650 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23244161 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23244169 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04004490 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03528409 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23244152 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23244171 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23244160 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23244184 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03270415 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23244166 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23244145 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23244164 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23244136 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23244182 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23244132 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23244157 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10507608 - -
          Affymetrix Mouse Gene 1.0 ST Array 10507611 - -
          Affymetrix Mouse Gene 1.0 ST Array 10507607 - -
          Affymetrix Mouse Gene 1.0 ST Array 10507610 - -
          Affymetrix Mouse Gene 1.0 ST Array 10507609 - -
          Affymetrix HT MG-430 PM Array Plate 1425616_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10507606 - -
          Mouse Genome 430 2.0 Array 1425616_a_at - -
          Mouse Genome 430 2.0 Array 1425616_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69216_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69216_at - -
          Affymetrix Mouse Clariom S Array PSR0400010043.mm.1 NM_001038998.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400010043.mm.1 NM_001038998.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400010043.mm.1 NM_024462.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400010043.mm.1 NM_024462.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400010043.mm.1 TC0400001327.mm.1 MTA TC
          Mouse Expression Set 430 1425616_PM_a_at - -
          Mouse Expression Set 430 1425616_a_at - -
          Mouse Genome 430A 2.0 Array 1425616_a_at - -
          Mouse Genome 430A 2.0 Array 1425616_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1425616_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1425616_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 913372 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3060441 - -
          Illumina mouse-6 v1.0 expression beadchip 3060441 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003060441 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2681604 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2681604 - -
          mouse-6 v1.1 (Illumina) scl25031.4.1_22-S - -
          Illumina Sentrix mouse-6.v1 bead array 3060441 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl25031.4.1_22-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3060441 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2681604 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2681604 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl25031.4.1_22-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline