Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00508361_m1
          See other 1810032O08RIK GE Assays ›
          Gene Symbol:
          1810032O08Rik
          Gene Name:
          RIKEN cDNA 1810032O08 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 116671660 - 116675798 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00508361_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1810032O08Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027819.1 3 - 4 366 66
          NR_027820.1 2 - 3 174 66
          NR_027821.1 3 - 4 366 66
          GenBank mRNA AK007675.1 - 3 - 4 367 66
          AK013681.1 - 2 - 3 166 66
          AK037979.1 - 3 - 4 358 66
          AK088915.1 - 3 - 4 349 66
          BC055707.1 - 3 - 4 352 66
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA NM_025472.1 - 358 Chr. - - [-] on Build 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001940.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 1810032O08Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2126448 AK037979.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2126448 AK088915.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2126448 BC055707.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2126448 NR_027819.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2126448 NR_027820.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2126444 AK037979.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2126444 BC055707.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2126444 NR_027821.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2126444 BC010208.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 867190 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 841526 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 608633 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P134755 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2126448 AK037979.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2126448 AK088915.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2126448 BC055707.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2126448 NR_027819.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2126448 NR_027820.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P117666 AK037979.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P117666 AK088915.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P117666 BC055707.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P117666 NR_027819.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P117666 NR_027820.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10382842 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382839 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382843 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382841 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382840 - -
          Affymetrix HT MG-430 PM Array Plate 1453344_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1436677_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10393166 - -
          Affymetrix Mouse Gene 1.1 ST Array 10382838 - -
          Mouse Genome 430 2.0 Array 1436677_PM_at - -
          Mouse Genome 430 2.0 Array 1453344_at - -
          Mouse Genome 430 2.0 Array 1436677_at - -
          Mouse Genome 430 2.0 Array 1453344_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66293_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66293_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100018442.mm.1 NR_027819.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018442.mm.1 NR_027819.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018442.mm.1 NR_027820.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018442.mm.1 NR_027820.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018442.mm.1 NR_027821.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018442.mm.1 NR_027821.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018442.mm.1 TC1100001940.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100018441.mm.1 NR_027819.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018441.mm.1 NR_027819.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018441.mm.1 NR_027820.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018441.mm.1 NR_027820.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018441.mm.1 NR_027821.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018441.mm.1 NR_027821.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018441.mm.1 TC1100001940.mm.1 MTA TC
          Mouse Expression Set 430 1436677_PM_at - -
          Mouse Expression Set 430 1436677_at - -
          Mouse Expression Set 430 1453344_at - -
          Mouse Expression Set 430 1453344_PM_at - -
          Mouse Genome 430A 2.0 Array 1436677_at - -
          Mouse Genome 430A 2.0 Array 1436677_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1436677_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1436677_at - -
          Affymetrix HT Mouse Genome 430B Array 1453344_at - -
          Affymetrix HT Mouse Genome 430B Array 1453344_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 751622 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 105720441 - -
          MouseWG-6 v2 Expression BeadChip 103830750 - -
          Illumina mouse-6 v1.0 expression beadchip 103830750 - -
          Illumina mouse-6 v1.0 expression beadchip 105720441 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1227976 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1230176 - -
          Illumina Sentrix mouse-6.v1 bead array 103830750 - -
          Illumina Sentrix mouse-6.v1 bead array 105720441 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 105720441 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 103830750 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1227976 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1230176 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline