Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00508398_g1
          See other 1700020L24RIK GE Assays ›
          Gene Symbol:
          1700020L24Rik
          Gene Name:
          RIKEN cDNA 1700020L24 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 83437694 - 83441235 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00508398_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700020L24Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025492.3 NP_079768.2 2 - 3 376 118
          GenBank mRNA AK005809.1 - 2 - 3 375 118
          AK006174.1 - 2 - 3 377 118
          BC152320.1 - 2 - 3 363 40136395 118

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001251.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 1700020L24Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27971501 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27971499 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27971504 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27971502 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27971503 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27971500 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P394921 NM_025492.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P394921 AK005809.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P394921 AK006174.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P394921 BC152320.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P394921 NM_025492.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P394921 AK005809.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P394921 AK006174.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P394921 BC152320.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 24803 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 787535 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 521948 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 68052 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P176918 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P394921 NM_025492.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P394921 AK005809.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P394921 AK006174.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P394921 BC152320.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P394921 NM_025492.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P394921 AK005809.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P394921 AK006174.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P394921 BC152320.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27971488 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27971494 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27971496 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27971498 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27971492 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27971491 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27971489 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27971486 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P176918 - -
          Whole Mouse Genome G4122A A_51_P394921 NM_025492.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P394921 AK005809.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P394921 AK006174.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P394921 BC152320.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P394921 NM_025492.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P394921 AK005809.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P394921 AK006174.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P394921 BC152320.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136759 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P176918 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09501877 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27971500 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27971501 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27971499 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27971503 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27971504 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27971502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27971453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27971500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27971465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27971440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27971502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27971449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27971438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27971467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27971436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27971434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27971447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27971480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27971489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27971442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27971466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27971435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27971498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27971504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27971483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27971583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27971427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27971486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27971508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27971492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27971494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27971475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27971477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27971496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27971505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27971499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27971437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27971429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27971457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27971509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27971471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27971433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27971482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27971474 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27971491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27971482 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27971479 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27971488 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27971492 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27971466 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27971460 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27971496 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27971504 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27971489 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27971459 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09501877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27971457 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27971477 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27971494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09501881 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27971471 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27971474 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27971505 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27971483 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27971461 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27971467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27971509 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27971480 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09163038 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27971499 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09468791 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27971502 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27971462 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27971463 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27971475 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10379686 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379688 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379687 - -
          Affymetrix HT MG-430 PM Array Plate 1453779_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10379685 - -
          Affymetrix Mouse Gene 1.1 ST Array 10389177 - -
          Mouse Genome 430 2.0 Array 1447544_at - -
          Mouse Genome 430 2.0 Array 1453779_PM_at - -
          Mouse Genome 430 2.0 Array 1453779_at - -
          Mouse Genome 430 2.0 Array 1447544_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66330_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66330_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100011758.mm.1 NM_025492.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011758.mm.1 NM_025492.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011758.mm.1 TC1100001251.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100011759.mm.1 NM_025492.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011759.mm.1 NM_025492.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011759.mm.1 TC1100001251.mm.1 MTA TC
          Mouse Expression Set 430 1453779_at - -
          Mouse Expression Set 430 1453779_PM_at - -
          Murine Genome U74v2 Set 169501_PM_r_at - -
          Murine Genome U74v2 Set 169501_r_at - -
          Affymetrix HT Mouse Genome 430B Array 1453779_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453779_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 379506 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5270592 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005270592 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2694101 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2694101 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2453145 - -
          mouse-6 v1.1 (Illumina) scl41137.3_142-S - -
          Illumina Sentrix mouse-6.v1 bead array 5270592 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl41137.3_142-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5270592 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2895240 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2895240 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline