Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00508436_m1
          See other TSFM GE Assays ›
          Gene Symbol:
          Tsfm
          Gene Name:
          Ts translation elongation factor, mitochondrial
          Gene Aliases:
          2310050B20Rik, 9430024O13Rik, EF-TS, EF-Tsmt
          Chromosome Location:
          Chr.10: 127022332 - 127030814 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00508436_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tsfm
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025537.3 NP_079813.1 5 - 6 588 74
          GenBank mRNA AJ277891.1 - 4 - 5 425 74
          AK009898.1 - 5 - 6 609 74
          AK012240.1 - 5 - 6 579 74
          AK018591.1 - 5 - 6 577 74
          AK169164.1 - 5 - 6 579 74
          AK180094.1 - 1 - 2 102 74
          BC034286.1 - 5 - 6 567 74
          BC051508.1 - 2 - 3 169 74
          BC057904.1 - 5 - 6 588 5012380 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000003093.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tsfm

          Panther Classification:

          Molecular Function -

          translation elongation factor

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          translational elongation
          mitochondrial translational elongation
          translation elongation factor activity
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27533592 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27533591 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27533589 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P235705 BC051508.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P235705 BC057904.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P235705 AK169164.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P235705 AK180094.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P235705 BC034286.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P235705 BC051508.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P235705 BC057904.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P235705 AK169164.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P235705 AK180094.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P235705 BC034286.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 214522 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 757823 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 392714 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 588334 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 49379 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 592256 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 112502 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P123149 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P235705 BC051508.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P235705 BC057904.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P235705 AK169164.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P235705 AK180094.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P235705 BC034286.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P235705 BC051508.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P235705 BC057904.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P235705 AK169164.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P235705 AK180094.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P235705 BC034286.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P235705 BC051508.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P235705 BC057904.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P235705 AK169164.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P235705 AK180094.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P235705 BC034286.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P235705 BC051508.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P235705 BC057904.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P235705 AK169164.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P235705 AK180094.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P235705 BC034286.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533607 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533610 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533593 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533605 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533597 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533619 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533579 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533631 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533600 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533616 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533634 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533602 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533584 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533585 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533612 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533620 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533624 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533594 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533625 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533580 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533586 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533635 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533628 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533606 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533622 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533590 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533595 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533614 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533589 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533591 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533598 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533630 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533581 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02244934 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P123149 - -
          Whole Mouse Genome G4122A A_51_P235705 BC051508.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P235705 BC057904.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P235705 AK169164.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P235705 AK180094.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P235705 BC034286.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P235705 BC051508.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P235705 BC057904.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P235705 AK169164.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P235705 AK180094.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P235705 BC034286.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132748 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P123149 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08941163 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08815423 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08815428 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08381736 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08208323 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27533592 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27533591 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27533589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533576 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533562 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533620 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533559 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533565 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533619 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533567 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533587 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533616 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533590 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08590478 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08962064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533600 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08702187 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08879279 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533605 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08714947 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08381735 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533610 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08870745 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533568 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08552505 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08552504 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533593 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533607 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533580 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533556 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533603 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533560 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533564 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533586 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08950395 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10373010 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373013 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373012 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373015 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373011 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373014 - -
          Affymetrix HT MG-430 PM Array Plate 1432170_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1452698_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10373009 - -
          Mouse Genome 430 2.0 Array 1452698_at - -
          Mouse Genome 430 2.0 Array 1432170_at - -
          Mouse Genome 430 2.0 Array 1432170_PM_at - -
          Mouse Genome 430 2.0 Array 1452698_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66399_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66399_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1000023019.mm.1 NM_025537.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023019.mm.1 NM_025537.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023019.mm.1 TC1000003093.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000023015.mm.1 NM_025537.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023015.mm.1 NM_025537.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023015.mm.1 TC1000003093.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000023014.mm.1 NM_025537.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023014.mm.1 NM_025537.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023014.mm.1 TC1000003093.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000023016.mm.1 NM_025537.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023016.mm.1 NM_025537.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023016.mm.1 TC1000003093.mm.1 MTA TC
          Mouse Expression Set 430 1452698_PM_at - -
          Mouse Expression Set 430 1452698_at - -
          Murine Genome U74v2 Set 97403_PM_at - -
          Murine Genome U74v2 Set 97403_at - -
          Murine Genome U74v2 Set 168185_i_at - -
          Murine Genome U74v2 Set 168185_PM_i_at - -
          Affymetrix HT Mouse Genome 430B Array 1452698_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1452698_at - -
          Affymetrix HT Mouse Genome 430B Array 1432170_at - -
          Affymetrix HT Mouse Genome 430B Array 1432170_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5220411 - -
          Illumina mouse-6 v1.0 expression beadchip 5220411 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005220411 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2640203 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2640203 - -
          mouse-6 v1.1 (Illumina) scl37403.6.1_51-S - -
          Illumina Sentrix mouse-6.v1 bead array 5220411 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl37403.6.1_51-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5220411 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2640203 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2640203 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl37403.6.1_51-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-knt4q:80/100.66.75.14:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0