Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00508510_m1
          See other TIMM50 GE Assays ›
          Gene Symbol:
          Timm50
          Gene Name:
          translocase of inner mitochondrial membrane 50
          Gene Aliases:
          2810403L02Rik, AU015082, TIM50L
          Chromosome Location:
          Chr.7: 28305516 - 28312046 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00508510_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Timm50
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025616.3 NP_079892.1 4 - 5 331 74
          GenBank mRNA AK008340.1 - 4 - 5 332 74
          AK008382.1 - 4 - 5 329 74
          AK010832.1 - 4 - 5 297 74
          AK012977.1 - 4 - 5 328 74
          AK088074.1 - 4 - 5 324 74
          AK167473.1 - 4 - 5 334 74
          AK178698.1 - 3 - 4 105 74
          AY551342.1 - 4 - 5 317 74
          BC010303.1 - 4 - 5 306 3597676 74
          BC037056.1 - 4 - 5 333 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700002693.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the BC010303.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Timm50

          Gene Ontology Categories:

          Function(s) Process(es)

          release of cytochrome c from mitochondria
          protein dephosphorylation
          transport
          mitochondrial membrane organization
          protein transport
          dephosphorylation
          peptidyl-tyrosine dephosphorylation
          phosphoprotein phosphatase activity
          protein serine/threonine phosphatase activity
          protein tyrosine phosphatase activity
          interleukin-2 receptor binding
          ribonucleoprotein complex binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25021122 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25021123 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25021124 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25021121 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P682990 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2127692 AY551342.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2127692 BC010303.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2127692 BC037056.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2127692 AK088074.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2127692 AK167473.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 136580 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 390100 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 198859 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 280193 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 354527 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 112058 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P176288 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2127692 AY551342.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2127692 BC010303.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2127692 BC037056.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2127692 AK088074.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2127692 AK167473.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P682990 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021079 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021130 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021082 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021112 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021108 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021116 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021111 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021122 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021075 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021126 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021089 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021086 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021129 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021125 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021131 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021083 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021133 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021113 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021114 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021110 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021109 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021123 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021120 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021105 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021087 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021119 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021107 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021115 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021080 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021127 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021124 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25021078 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01489803 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P176288 - -
          Whole Mouse Genome G4122A A_52_P682990 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103929 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P176288 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06614672 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06614677 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06614669 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25021121 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25021124 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25021123 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25021122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021105 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021089 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021100 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06123939 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06099641 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021094 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021131 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06278951 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021133 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06050089 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06280847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021095 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021111 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06099624 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06614669 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021113 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021115 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06452797 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021093 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021120 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021098 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06614672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021083 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021104 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06280858 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06614677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06099640 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25021119 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10561392 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561398 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561395 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561399 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561389 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561391 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561397 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561394 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561393 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561400 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561390 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561396 - -
          Affymetrix HT MG-430 PM Array Plate 1455456_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1441819_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1459830_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10561388 - -
          Mouse Genome 430 2.0 Array 1441819_x_at - -
          Mouse Genome 430 2.0 Array 1455456_a_at - -
          Mouse Genome 430 2.0 Array 1459830_PM_at - -
          Mouse Genome 430 2.0 Array 1441819_PM_x_at - -
          Mouse Genome 430 2.0 Array 1459830_at - -
          Mouse Genome 430 2.0 Array 1455456_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66525_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66525_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700023514.mm.1 TC0700002693.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700023514.mm.1 NM_025616.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700023514.mm.1 NM_025616.3 Refseq
          Mouse Expression Set 430 1455456_a_at - -
          Mouse Expression Set 430 1455456_PM_a_at - -
          Mouse Expression Set 430 1459830_at - -
          Mouse Expression Set 430 1459830_PM_at - -
          Murine Genome U74v2 Set 168614_at - -
          Murine Genome U74v2 Set 168614_PM_at - -
          Mouse Genome 430A 2.0 Array 1455456_a_at - -
          Mouse Genome 430A 2.0 Array 1455456_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1455456_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1455456_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1459830_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1441819_PM_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1441819_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1459830_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 918832 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1500139 - -
          Illumina mouse-6 v1.0 expression beadchip 1500139 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001500139 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1241845 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1241845 - -
          mouse-6 v1.1 (Illumina) scl31584.11.1_18-S - -
          Illumina Sentrix mouse-6.v1 bead array 1500139 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl31584.11.1_18-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1500139 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1241845 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1241845 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl31584.11.1_18-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline