Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00508985_m1
          See other BPIFA5 GE Assays ›
          Gene Symbol:
          Bpifa5
          Gene Name:
          BPI fold containing family A, member 5
          Gene Aliases:
          2310021H06Rik, 2310074B19Rik, Pluncl, Splunc5, Tpl
          Chromosome Location:
          Chr.2: 154162607 - 154168446 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00508985_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Bpifa5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025990.4 NP_080266.2 7 - 8 837 139
          GenBank mRNA AK009329.1 - 7 - 8 833 139
          AK009441.1 - 7 - 8 839 139
          AK009531.1 - 7 - 8 833 139
          AK009562.1 - 7 - 8 833 139
          AK009580.1 - 7 - 8 837 139
          AK009629.1 - 7 - 8 836 139
          AK009695.1 - 7 - 8 834 139
          AK009803.1 - 7 - 8 835 139
          AK009835.1 - 7 - 8 139
          AK010012.1 - 7 - 8 837 139
          AK010051.1 - 7 - 8 834 139
          AK010115.1 - 7 - 8 833 139
          AK010143.1 - 6 - 7 779 139
          AK010145.1 - 7 - 8 837 139
          AK010158.1 - 6 - 7 754 139
          BC115526.1 - 7 - 8 816 40101263 139
          BC115527.1 - 7 - 8 813 40101264 139

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200002336.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Transporter 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Bpifa5

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          lipid binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P219722 AK010145.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P219722 AK010115.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P219722 AK010143.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P219722 AK010158.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P219722 AK010159.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P219722 AK010145.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P219722 AK010115.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P219722 AK010143.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P219722 AK010158.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P219722 AK010159.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 613453 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 569978 AK038554.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 302554 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 552870 BC050987.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 552870 BC100365.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 359788 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 121797 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P139413 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P219722 AK010145.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P219722 AK010115.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P219722 AK010143.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P219722 AK010158.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P219722 AK010159.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P219722 AK010145.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P219722 AK010115.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P219722 AK010143.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P219722 AK010158.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P219722 AK010159.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21759048 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21759044 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21759039 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21759060 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21759021 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21759054 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21759040 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21759045 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21759066 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21759057 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21759041 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21759020 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21759053 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21759061 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21759034 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21759052 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21759023 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21759058 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21759029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21759025 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21759032 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P139413 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04746284 - -
          Whole Mouse Genome G4122A A_51_P219722 AK010145.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P219722 AK010115.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P219722 AK010143.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P219722 AK010158.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P219722 AK010159.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P219722 AK010145.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P219722 AK010115.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P219722 AK010143.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P219722 AK010158.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P219722 AK010159.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105860 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117468 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P139413 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01970414 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01970418 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01754570 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01970410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21759032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21759006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21759044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21759068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21759103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21759091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21759067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21759083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21759015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21759075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21759019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21759111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21759017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21759010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21759107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21759110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21759096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21759092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21759048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21759054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21759077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21759041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21759066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21759085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21759106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21759040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21759070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21759073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21759057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21759072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21759025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21759009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21759078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21759071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21759081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21759045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21759090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21759069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21759094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21759088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21759105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21759011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21758994 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01824779 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21759073 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21759064 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01608199 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21759095 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21759077 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21759029 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21759044 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21759081 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21759083 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21759010 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21759085 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21759011 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21759052 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21759034 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21759048 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01292859 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21759066 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21759020 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21759027 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21759078 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21759075 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21759061 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02075736 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01970412 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01325969 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21759067 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21759068 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21759054 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01325974 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21759014 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21759090 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21759096 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21759092 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01543918 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21759072 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02082774 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01479186 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01292864 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01970418 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01970410 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01754569 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01978577 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01292865 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01325990 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21759069 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01325987 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21759094 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21759038 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21759015 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01754571 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21759021 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10477492 - -
          Affymetrix Mouse Gene 1.0 ST Array 10477488 - -
          Affymetrix Mouse Gene 1.0 ST Array 10477491 - -
          Affymetrix Mouse Gene 1.0 ST Array 10477494 - -
          Affymetrix Mouse Gene 1.0 ST Array 10477487 - -
          Affymetrix Mouse Gene 1.0 ST Array 10477490 - -
          Affymetrix Mouse Gene 1.0 ST Array 10477489 - -
          Affymetrix Mouse Gene 1.0 ST Array 10477486 - -
          Affymetrix Mouse Gene 1.0 ST Array 10477493 - -
          Affymetrix HT MG-430 PM Array Plate 1429342_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10477485 - -
          Mouse Genome 430 2.0 Array 1429342_PM_s_at - -
          Mouse Genome 430 2.0 Array 1429342_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67135_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67135_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200018496.mm.1 NM_025990.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200018496.mm.1 NM_025990.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200018496.mm.1 TC0200002336.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200018497.mm.1 NM_025990.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200018497.mm.1 NM_025990.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200018497.mm.1 TC0200002336.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200018498.mm.1 NM_025990.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200018498.mm.1 NM_025990.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200018498.mm.1 TC0200002336.mm.1 MTA TC
          Mouse Expression Set 430 1429342_PM_s_at - -
          Mouse Expression Set 430 1429342_s_at - -
          Murine Genome U74v2 Set 171517_at - -
          Murine Genome U74v2 Set 171517_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429342_PM_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1429342_s_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6520132 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006520132 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2653590 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2653590 - -
          mouse-6 v1.1 (Illumina) scl067135.5_1-S - -
          Illumina Sentrix mouse-6.v1 bead array 6520132 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl067135.5_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6520132 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2653590 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2653590 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline