Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00509126_m1
          See other IFT22 GE Assays ›
          Gene Symbol:
          Ift22
          Gene Name:
          intraflagellar transport 22
          Gene Aliases:
          3110017O03Rik, AI835745, AI847427, Rabl5
          Chromosome Location:
          Chr.5: 136908150 - 136914509 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00509126_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ift22
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026073.3 NP_080349.1 4 - 5 493 99
          GenBank mRNA AK005820.1 - 4 - 5 453 99
          BC009150.1 - 4 - 5 493 2651447 99

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500001610.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ift22

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular protein transport
          small GTPase mediated signal transduction
          Rab protein signal transduction
          intraciliary transport
          nucleotide binding
          molecular_function
          GTPase activity
          GTP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24123651 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24123653 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24123655 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24123652 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24123654 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P495077 BC009150.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P495077 BC009150.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 787108 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 93149 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 326229 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 75542 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 62480 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P177832 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P495077 BC009150.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P495077 BC009150.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24123592 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24123596 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24123611 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24123613 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24123590 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24123607 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24123601 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24123594 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24123598 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24123609 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24123605 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P177832 - -
          Whole Mouse Genome G4122A A_51_P495077 BC009150.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P495077 BC009150.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126957 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P177832 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04453939 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04453938 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04453936 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24123651 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24123654 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24123655 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24123652 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24123653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123656 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123596 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04123081 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04944210 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123584 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123622 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123630 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123588 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123636 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04683521 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123669 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123601 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123607 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123646 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04944212 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123613 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123583 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123664 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04773257 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123649 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123648 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04274745 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04870296 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123661 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123626 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04865509 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04566324 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123642 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04235093 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04865511 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04243398 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04420225 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123585 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04151083 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123659 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04813135 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04870293 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123581 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123605 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123643 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123612 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04870291 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123625 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04769289 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04453939 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04104182 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04865506 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04211215 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04957880 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123635 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123656 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123631 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123611 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04683520 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123593 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04336421 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04897091 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123658 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123620 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04897090 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10526505 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526506 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526507 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526503 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526504 - -
          Affymetrix HT MG-430 PM Array Plate 1424575_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10526502 - -
          Mouse Genome 430 2.0 Array 1424575_PM_at - -
          Mouse Genome 430 2.0 Array 1424575_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67286_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67286_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0500014546.mm.1 TC0500001610.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500014546.mm.1 NM_026073.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500014546.mm.1 NM_026073.3 Refseq
          Mouse Expression Set 430 1424575_PM_at - -
          Mouse Expression Set 430 1424575_at - -
          Mouse Genome 430A 2.0 Array 1424575_PM_at - -
          Mouse Genome 430A 2.0 Array 1424575_at - -
          Affymetrix HT Mouse Genome 430A Array 1424575_at - -
          Affymetrix HT Mouse Genome 430A Array 1424575_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 540848 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 770368 - -
          Illumina mouse-6 v1.0 expression beadchip 770368 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000770368 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1218444 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1218444 - -
          mouse-6 v1.1 (Illumina) scl27111.3.1_10-S - -
          Illumina Sentrix mouse-6.v1 bead array 770368 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl27111.3.1_10-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 770368 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2637398 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2637398 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl27111.3.1_10-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline