Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00509129_m1
          See other IQCF4 GE Assays ›
          Gene Symbol:
          Iqcf4
          Gene Name:
          IQ motif containing F4
          Gene Aliases:
          1700042N06Rik, mtIQ1
          Chromosome Location:
          Chr.9: 106568319 - 106570967 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00509129_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Iqcf4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026090.2 NP_080366.2 2 - 3 267 106
          GenBank mRNA AK006688.1 - 2 - 3 266 106
          BC048652.1 - 2 - 3 272 6772797 106
          BC061014.1 - 2 - 3 267 6770556 106
          DQ153246.1 - 2 - 3 296 106

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900003014.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Iqcf4

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2053823 BC048652.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2053823 BC061014.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2053823 AK006688.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2053823 DQ153246.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2053823 NM_026090.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2053824 BC061014.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2053824 AK006688.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2053824 DQ153246.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2053824 NM_026090.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 853277 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 351223 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 790337 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 171846 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P173629 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2053823 BC048652.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2053823 BC061014.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2053823 AK006688.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2053823 DQ153246.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2053823 NM_026090.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2053824 BC061014.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2053824 AK006688.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2053824 DQ153246.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2053824 NM_026090.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26804041 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26804046 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26804054 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26804044 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26804056 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26804040 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26804052 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26804050 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26804057 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26804068 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26804070 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26804064 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26804063 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26804060 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26804038 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26804065 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P173629 - -
          Whole Mouse Genome G4122A A_51_P513764 BC048652.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P513764 BC061014.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P513764 AK006688.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P513764 DQ153246.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P513764 NM_026090.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131564 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113234 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P173629 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07946664 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07946673 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07946657 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07946672 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07946671 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07946659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26804040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26804009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26804044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26804039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26804006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26804068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26804026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26804014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26804063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26804022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26804012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26804005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26804052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26804054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26804050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26804076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26804056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26804070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26804058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26804010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26804047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26804018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26804077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26804003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26804016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26804037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26804000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26804021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26804020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26804015 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804057 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804060 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804058 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804050 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07946671 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804040 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804063 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08067008 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08086415 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07946666 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07946657 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804054 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804061 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804047 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07884035 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07807955 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804070 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10596520 - -
          Affymetrix Mouse Gene 1.0 ST Array 10596519 - -
          Affymetrix Mouse Gene 1.0 ST Array 10596518 - -
          Affymetrix HT MG-430 PM Array Plate 1429577_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10596517 - -
          Mouse Genome 430 2.0 Array 1429577_at - -
          Mouse Genome 430 2.0 Array 1429577_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67320_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67320_at - -
          Affymetrix Mouse Clariom S Array PSR0900025064.mm.1 NM_026090.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025064.mm.1 NM_026090.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025064.mm.1 TC0900003014.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900025065.mm.1 NM_026090.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025065.mm.1 NM_026090.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025065.mm.1 TC0900003014.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900025063.mm.1 NM_026090.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025063.mm.1 NM_026090.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025063.mm.1 TC0900003014.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900025066.mm.1 NM_026090.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025066.mm.1 NM_026090.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025066.mm.1 TC0900003014.mm.1 MTA TC
          Mouse Expression Set 430 1429577_at - -
          Mouse Expression Set 430 1429577_PM_at - -
          Murine Genome U74v2 Set 167370_at - -
          Murine Genome U74v2 Set 167370_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429577_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429577_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 673549 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 840524 - -
          Illumina mouse-6 v1.0 expression beadchip 840524 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000840524 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2677261 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2677261 - -
          mouse-6 v1.1 (Illumina) scl35391.3.1_2-S - -
          Illumina Sentrix mouse-6.v1 bead array 840524 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl35391.3.1_2-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 840524 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2677261 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2677261 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline