Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00509340_m1
          See other GIN1 GE Assays ›
          Gene Symbol:
          Gin1
          Gene Name:
          gypsy retrotransposon integrase 1
          Gene Aliases:
          4930429M06Rik, 8430411H10Rik, Gin-1, Zh2c2
          Chromosome Location:
          Chr.1: 97770168 - 97793710 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00509340_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gin1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026250.2 NP_080526.2 7 - 8 1364 68
          GenBank mRNA AK014287.1 - 7 - 8 1371 68
          AK015243.1 - 7 - 8 1343 68
          AK090002.1 - 7 - 8 1082 68
          AK169361.1 - 7 - 8 1327 68
          BC033572.1 - 6 - 7 1309 5067692 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100000971.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gin1

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA integration
          nucleic acid binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20472807 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20472804 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20472806 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20472805 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P392509 AK018404.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P392509 AK084620.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P392509 AK018404.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P392509 AK084620.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2078895 AK014287.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2078895 NM_026250.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2078895 AK015243.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 875321 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 434199 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 18013 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 970840 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 903335 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 334642 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 791118 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 561896 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 825288 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 873869 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 493316 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 305071 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 163419 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 130623 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P143278 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P175896 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P392509 AK018404.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P392509 AK084620.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P392509 AK018404.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P392509 AK084620.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2078895 AK014287.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2078895 NM_026250.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2078895 AK015243.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P365932 BC033572.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P365932 AK090002.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P365932 AK169361.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P365932 AK014287.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P365932 NM_026250.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20472816 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20472818 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20472814 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20472808 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20472805 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20472813 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20472806 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20472804 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20472811 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04200607 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04200554 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P175896 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P143278 - -
          Whole Mouse Genome G4122A A_52_P392509 AK018404.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P392509 AK084620.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P392509 AK018404.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P392509 AK084620.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P365932 BC033572.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P365932 AK090002.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P365932 AK169361.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P365932 AK014287.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P365932 NM_026250.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116675 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P143278 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P175896 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00954122 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00954128 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00369978 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00954124 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20472806 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20472805 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20472804 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20472807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20472906 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20472843 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20472806 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20472840 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20472823 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20472815 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00992787 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20472814 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20472805 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20472817 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00369979 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20472811 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00979557 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00954122 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20472845 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00968138 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00241969 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00954126 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20472804 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20472825 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20472808 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20472844 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20472835 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20472848 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20472826 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20472813 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20472837 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20472829 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00954121 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10348934 - -
          Affymetrix Mouse Gene 1.0 ST Array 10348941 - -
          Affymetrix Mouse Gene 1.0 ST Array 10348937 - -
          Affymetrix Mouse Gene 1.0 ST Array 10348935 - -
          Affymetrix Mouse Gene 1.0 ST Array 10348940 - -
          Affymetrix Mouse Gene 1.0 ST Array 10348939 - -
          Affymetrix Mouse Gene 1.0 ST Array 10348938 - -
          Affymetrix Mouse Gene 1.0 ST Array 10348936 - -
          Affymetrix Mouse Gene 1.0 ST Array 10348933 - -
          Affymetrix HT MG-430 PM Array Plate 1429118_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1429728_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10348932 - -
          Mouse Genome 430 2.0 Array 1429118_a_at - -
          Mouse Genome 430 2.0 Array 1429118_PM_a_at - -
          Mouse Genome 430 2.0 Array 1429728_PM_at - -
          Mouse Genome 430 2.0 Array 1429728_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 252876_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 252876_at - -
          Affymetrix Mouse Clariom S Array PSR0100008207.mm.1 NM_026250.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100008207.mm.1 TC0100000971.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100008208.mm.1 NM_026250.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100008208.mm.1 TC0100000971.mm.1 MTA TC
          Mouse Expression Set 430 1429118_PM_a_at - -
          Mouse Expression Set 430 1429118_a_at - -
          Mouse Expression Set 430 1429728_at - -
          Mouse Expression Set 430 1429728_PM_at - -
          Murine Genome U74v2 Set 166315_PM_at - -
          Murine Genome U74v2 Set 166315_at - -
          Affymetrix HT Mouse Genome 430B Array 1429728_at - -
          Affymetrix HT Mouse Genome 430B Array 1429118_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1429728_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429118_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 507110 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104810735 - -
          MouseWG-6 v2 Expression BeadChip 104560338 - -
          MouseWG-6 v2 Expression BeadChip 104760605 - -
          Illumina mouse-6 v1.0 expression beadchip 4210019 - -
          Illumina mouse-6 v1.0 expression beadchip 104810735 - -
          Illumina mouse-6 v1.0 expression beadchip 104760605 - -
          Illumina mouse-6 v1.0 expression beadchip 104560338 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004210019 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2707273 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1229362 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2493476 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2707273 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1245008 - -
          mouse-6 v1.1 (Illumina) scl00252876.2_224-S - -
          mouse-6 v1.1 (Illumina) scl17607.9_409-S - -
          Illumina Sentrix mouse-6.v1 bead array 104810735 - -
          Illumina Sentrix mouse-6.v1 bead array 4210019 - -
          Illumina Sentrix mouse-6.v1 bead array 104760605 - -
          Illumina Sentrix mouse-6.v1 bead array 104560338 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00252876.2_224-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104560338 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104810735 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4210019 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104760605 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2707273 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1229362 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1245008 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2707273 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline