Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00509423_m1
          See other 4930522H14RIK GE Assays ›
          Gene Symbol:
          4930522H14Rik
          Gene Name:
          RIKEN cDNA 4930522H14 gene
          Gene Aliases:
          1700007J23Rik
          Chromosome Location:
          Chr.4: 109505332 - 109531297 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00509423_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930522H14Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001199090.1 NP_001186019.1 3 - 4 358 108
          NM_026291.3 NP_080567.1 3 - 4 335 108
          GenBank mRNA AK005726.1 - 3 - 4 359 108
          AK015869.1 - 3 - 4 277 108
          AK019777.1 - 3 - 4 336 108
          BC125547.1 - 3 - 4 295 40130062 108
          BC125549.1 - 3 - 4 296 40130064 108

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400003269.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 4930522H14Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952284 AK015869.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952284 AK019777.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952284 BC125547.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952284 BC125549.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952284 NM_001199090.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 407820 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 676164 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 922818 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 256174 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 596971 AK051595.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 596971 AK164723.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 678111 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 795836 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 642562 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 764155 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 35126 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 108384 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P155667 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P163797 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1952284 AK015869.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1952284 AK019777.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1952284 BC125547.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1952284 BC125549.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1952284 NM_001199090.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23201534 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23201542 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23201545 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23201539 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23201552 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23201519 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23201535 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23201525 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23201527 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23201541 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23201556 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23201523 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23201537 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23201551 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23201531 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23201553 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23201557 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23201526 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23201521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23201528 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23201514 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23201517 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23201547 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23201516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23201546 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23201549 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P155667 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P163797 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05356486 - -
          Whole Mouse Genome G4122A A_51_P135607 AK015869.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P135607 AK019777.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P135607 BC125547.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P135607 BC125549.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P135607 NM_001199090.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107040 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P155667 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P163797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23201479 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201556 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201420 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201453 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04096332 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201527 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03721222 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201407 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201513 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201508 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03809830 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201546 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201454 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03606125 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201559 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201414 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201552 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201447 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201515 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03527295 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201519 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03703062 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201511 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201509 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201443 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201441 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03690680 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04096330 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201540 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03333244 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201531 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03690678 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201528 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201497 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201512 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201545 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201523 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201491 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201536 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201564 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201498 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201490 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201533 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201415 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201507 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201448 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201493 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201539 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201500 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201409 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201445 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201521 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201541 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201417 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201411 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201551 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201496 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03735728 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201553 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03690675 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201504 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201526 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201562 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03270470 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201557 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201538 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03324433 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201534 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201408 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23201450 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10515081 - -
          Affymetrix Mouse Gene 1.0 ST Array 10515085 - -
          Affymetrix Mouse Gene 1.0 ST Array 10515082 - -
          Affymetrix Mouse Gene 1.0 ST Array 10515083 - -
          Affymetrix Mouse Gene 1.0 ST Array 10515084 - -
          Affymetrix HT MG-430 PM Array Plate 1431104_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10515080 - -
          Mouse Genome 430 2.0 Array 1431104_at - -
          Mouse Genome 430 2.0 Array 1431104_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67646_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67646_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400026050.mm.1 TC0400003269.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400026050.mm.1 NM_001199090.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400026050.mm.1 NM_001199090.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400026050.mm.1 NM_026291.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400026050.mm.1 NM_026291.3 Refseq
          Mouse Expression Set 430 1431104_at - -
          Mouse Expression Set 430 1431104_PM_at - -
          Murine Genome U74v2 Set 165857_PM_at - -
          Murine Genome U74v2 Set 165857_at - -
          Affymetrix HT Mouse Genome 430B Array 1431104_at - -
          Affymetrix HT Mouse Genome 430B Array 1431104_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 905568 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2360110 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002360110 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2720901 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2720901 - -
          mouse-6 v1.1 (Illumina) scl23994.6.1_16-S - -
          Illumina Sentrix mouse-6.v1 bead array 2360110 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl23994.6.1_16-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2360110 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2720901 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2720901 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline