Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00509436_m1
          See other CEP83OS GE Assays ›
          Gene Symbol:
          Cep83os
          Gene Name:
          centrosomal protein 83, opposite strand
          Gene Aliases:
          4932415G12Rik, Ccdc41os1
          Chromosome Location:
          Chr.10: 94673493 - 94688613 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00509436_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cep83os
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_015524.1 1 - 2 378 93
          GenBank mRNA AK015136.1 - 1 - 2 292 93
          AK016525.1 - 1 - 2 337 93
          AK085032.1 - 1 - 2 343 93
          AK144701.1 - 1 - 2 340 93
          AK154718.1 - 1 - 2 310 93
          AK155868.1 - 1 - 2 310 93
          BC067070.1 - 1 - 2 323 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000002713.mm.1

          Set Membership:

          Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 896219 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 628591 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 278881 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 63624 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 182923 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 647037 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 216119 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 555066 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 972023 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 757011 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 102469 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P105565 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P171934 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P171934 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07156500 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P105565 - -
          Whole Mouse Genome G4122A A_52_P279508 AK034425.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P152479 BC067070.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P152479 BC021358.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P152479 BC061482.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P152479 AK155868.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P152479 AK154718.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P564353 BC067070.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P564353 BC061482.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P564353 AK155868.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P564353 AK154718.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P564353 AK085032.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P105565 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P171934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27360654 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27360675 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27360643 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27360733 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08657437 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27360652 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27360657 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27360729 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27360682 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08551734 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27360731 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27360648 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27360678 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27360637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27360640 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27360686 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27360646 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27360641 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27360677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27360680 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27360659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08657443 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27360679 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27360730 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27360684 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1428376_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1428375_PM_at - -
          Mouse Genome 430 2.0 Array 1428376_PM_at - -
          Mouse Genome 430 2.0 Array 1428375_PM_at - -
          Mouse Genome 430 2.0 Array 1428375_at - -
          Mouse Genome 430 2.0 Array 1428376_at - -
          Affymetrix Mouse Clariom S Array PSR1000020850.mm.1 NR_015524.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000020850.mm.1 NR_015524.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000020850.mm.1 TC1000002713.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000020849.mm.1 NR_015524.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000020849.mm.1 NR_015524.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000020849.mm.1 TC1000002713.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000020845.mm.1 NR_015524.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000020845.mm.1 NR_015524.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000020845.mm.1 TC1000002713.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000020846.mm.1 NR_015524.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000020846.mm.1 NR_015524.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000020846.mm.1 TC1000002713.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000020839.mm.1 NR_015524.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000020839.mm.1 NR_015524.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000020839.mm.1 TC1000002713.mm.1 MTA TC
          Mouse Expression Set 430 1428375_PM_at - -
          Mouse Expression Set 430 1428375_at - -
          Mouse Expression Set 430 1428376_PM_at - -
          Mouse Expression Set 430 1428376_at - -
          Murine Genome U74v2 Set 168754_PM_i_at - -
          Murine Genome U74v2 Set 168754_i_at - -
          Affymetrix HT Mouse Genome 430B Array 1428375_at - -
          Affymetrix HT Mouse Genome 430B Array 1428376_at - -
          Affymetrix HT Mouse Genome 430B Array 1428375_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1428376_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 687684 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101570053 - -
          MouseWG-6 v2 Expression BeadChip 6900152 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline