Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00509473_m1
          See other CCDC130 GE Assays ›
          Gene Symbol:
          Ccdc130
          Gene Name:
          coiled-coil domain containing 130
          Gene Aliases:
          4930527D15Rik, C77430
          Chromosome Location:
          Chr.8: 84257767 - 84270458 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00509473_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccdc130
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001294281.1 NP_001281210.1 5 - 6 497 64
          NM_026350.3 NP_080626.1 5 - 6 497 64
          GenBank mRNA AK015911.1 - 5 - 6 456 64
          BC021321.1 - 4 - 5 453 5041100 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800002580.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ccdc130

          Panther Classification:

          Molecular Function -

          RNA splicing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26019094 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26019095 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26019096 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2404223 BC021321.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2404223 NM_026350.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2404223 AK015911.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2404223 BC008614.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 708587 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 218442 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 104452 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 499051 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 424306 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 949103 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 536297 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 310420 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 8745 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P118479 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P163458 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P168376 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2404223 BC021321.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2404223 NM_026350.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2404223 AK015911.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2404223 BC008614.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26019105 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26019099 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26019089 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26019045 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26019050 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26019101 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26019058 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26019056 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26019059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26019096 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26019093 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26019041 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26019052 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26019111 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26019026 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26019039 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26019024 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26019107 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26019042 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26019094 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26019100 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26019030 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26019109 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26019091 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26019110 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26019027 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26019106 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26019103 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26019046 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26019033 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26019047 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26019032 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26019023 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26019097 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26019022 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26019054 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26019036 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P118479 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01783256 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P168376 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P163458 - -
          Whole Mouse Genome G4122A A_52_P439932 BC021321.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P439932 NM_026350.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P439932 AK003379.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P439932 AK015911.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P439932 AK218503.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124003 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P168376 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P163458 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P118479 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07148376 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07187945 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07187938 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06669446 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07187936 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06669449 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26019095 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26019094 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26019096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26019065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019105 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019032 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07187942 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019021 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019093 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019047 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07155838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019091 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07155837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019046 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019066 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019033 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019050 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06974346 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07094210 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019082 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019077 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019022 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019055 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019054 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019058 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07186989 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019061 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019085 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07187952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019048 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06905466 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019109 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019030 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019024 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06806243 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019106 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06782834 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019052 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019045 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019111 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019098 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07141825 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019042 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06738104 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019034 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06820397 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07424506 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019100 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019063 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26019026 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10580177 - -
          Affymetrix Mouse Gene 1.0 ST Array 10580173 - -
          Affymetrix Mouse Gene 1.0 ST Array 10580180 - -
          Affymetrix Mouse Gene 1.0 ST Array 10580174 - -
          Affymetrix Mouse Gene 1.0 ST Array 10580179 - -
          Affymetrix Mouse Gene 1.0 ST Array 10580175 - -
          Affymetrix Mouse Gene 1.0 ST Array 10580178 - -
          Affymetrix Mouse Gene 1.0 ST Array 10580171 - -
          Affymetrix Mouse Gene 1.0 ST Array 10580172 - -
          Affymetrix Mouse Gene 1.0 ST Array 10580170 - -
          Affymetrix Mouse Gene 1.0 ST Array 10580176 - -
          Affymetrix HT MG-430 PM Array Plate 1440100_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1452523_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10580169 - -
          Mouse Genome 430 2.0 Array 1440100_at - -
          Mouse Genome 430 2.0 Array 1452523_PM_a_at - -
          Mouse Genome 430 2.0 Array 1440100_PM_at - -
          Mouse Genome 430 2.0 Array 1452523_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67736_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67736_at - -
          Affymetrix Mouse Clariom S Array PSR0800019683.mm.1 NM_026350.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800019683.mm.1 TC0800002580.mm.1 MTA TC
          Mouse Expression Set 430 1452523_PM_a_at - -
          Mouse Expression Set 430 1452523_a_at - -
          Mouse Expression Set 430 1440100_at - -
          Mouse Expression Set 430 1440100_PM_at - -
          Murine Genome U74v2 Set 104485_at - -
          Murine Genome U74v2 Set 104485_PM_at - -
          Murine Genome U74v2 Set 171575_PM_at - -
          Murine Genome U74v2 Set 171575_at - -
          Mouse Genome 430A 2.0 Array 1452523_a_at - -
          Mouse Genome 430A 2.0 Array 1452523_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1452523_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1452523_PM_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1440100_at - -
          Affymetrix HT Mouse Genome 430B Array 1440100_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 921222 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4050138 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004050138 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2756733 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2756733 - -
          mouse-6 v1.1 (Illumina) scl34564.9_192-S - -
          Illumina Sentrix mouse-6.v1 bead array 4050138 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl34564.9_192-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4050138 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2756733 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2756733 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl34564.9_192-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline