Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00509578_s1
          See other TEX13A GE Assays ›
          Gene Symbol:
          Tex13a
          Gene Name:
          testis expressed 13A
          Gene Aliases:
          1700025D03Rik
          Chromosome Location:
          Chr.X: 138208158 - 138209591 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00509578_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tex13a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026469.2 NP_080745.2 2 776 88
          GenBank mRNA AK006327.1 - 1 56 88

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tex13a

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P393014 NM_026469.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P393014 AK006327.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P393014 NM_026469.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P393014 AK006327.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P393014 NM_026469.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P393014 AK006327.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P393014 NM_026469.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P393014 AK006327.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32688872 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32688846 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32688865 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32688844 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32688876 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32688857 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32688873 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32688851 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32688862 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32688860 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32688856 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32688864 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32688859 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32688849 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32688871 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32688879 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32688863 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32688878 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32688867 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32688858 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32688852 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32688854 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32688870 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32688869 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32688861 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32688848 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32688842 - -
          Whole Mouse Genome G4122A A_51_P393014 NM_026469.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P393014 AK006327.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P393014 NM_026469.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P393014 AK006327.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P37613 AK006452.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P37613 NM_026469.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P37613 AK006327.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137042 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15082166 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15082162 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14927428 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14617693 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14754689 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15082159 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14754688 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15082158 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15082160 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14927426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32688870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32688882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32688876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32688868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32688861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32688896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32688844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32688863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32688873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32688848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32688869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32688887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32688846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32688879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32688858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32688880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32688857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32688852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32688860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32688862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32688842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32688865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32688889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32688891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32688898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32688851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32688885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32688884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32688894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32688849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32688872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32688854 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32688864 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32688852 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14754687 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32688860 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32688886 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15133195 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32688865 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32688882 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32688887 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32688858 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32688851 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14927427 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32688876 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32688862 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32688859 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32688881 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14845689 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32688846 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14927426 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32688844 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32688879 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32688873 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32688854 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32688891 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32688848 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32688863 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32688869 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32688868 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15133193 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32688849 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15082168 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32688880 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32688870 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14845694 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10606927 - -
          Affymetrix HT MG-430 PM Array Plate 1431531_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10606926 - -
          Mouse Genome 430 2.0 Array 1431531_PM_at - -
          Mouse Genome 430 2.0 Array 1431531_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67944_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67944_PM_at - -
          Mouse Expression Set 430 1431531_at - -
          Mouse Expression Set 430 1431531_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431531_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431531_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3840193 - -
          Illumina mouse-6 v1.0 expression beadchip 102450575 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003840193 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1255543 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1255543 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1222335 - -
          mouse-6 v1.1 (Illumina) scl53798.2.109_205-S - -
          Illumina Sentrix mouse-6.v1 bead array 3840193 - -
          Illumina Sentrix mouse-6.v1 bead array 102450575 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl53798.2.109_205-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3840193 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102450575 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1255543 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1222335 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1255543 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline