Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00509675_s1
          See other 5031426D15RIK GE Assays ›
          Gene Symbol:
          5031426D15Rik
          Gene Name:
          RIKEN cDNA 5031426D15 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 6922702 - 6928722 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00509675_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 5031426D15Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027890.1 1 1853 141
          GenBank mRNA AK019879.1 - 1 1852 141
          AK032294.1 - 1 41 141

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200002910.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Whole Mouse Genome G4122A A_51_P149579 AK032294.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P149579 AK019879.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P149579 NR_027890.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1431248_PM_at - -
          Mouse Genome 430 2.0 Array 1431248_PM_at - -
          Mouse Genome 430 2.0 Array 1431248_at - -
          Affymetrix Mouse Clariom S Array PSR0200024146.mm.1 NR_027890.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200024146.mm.1 NR_027890.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200024146.mm.1 TC0200002910.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200024147.mm.1 NR_027890.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200024147.mm.1 NR_027890.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200024147.mm.1 TC0200002910.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200024148.mm.1 NR_027890.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200024148.mm.1 NR_027890.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200024148.mm.1 TC0200002910.mm.1 MTA TC
          Mouse Expression Set 430 1431248_PM_at - -
          Mouse Expression Set 430 1431248_at - -
          Affymetrix HT Mouse Genome 430B Array 1431248_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431248_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 396630 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0