Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00510008_m1
          See other ZFP688 GE Assays ›
          Gene Symbol:
          Zfp688
          Gene Name:
          zinc finger protein 688
          Gene Aliases:
          2810407K09Rik, AI326029
          Chromosome Location:
          Chr.7: 127418966 - 127423640 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00510008_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp688
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026999.4 NP_081275.3 2 - 3 340 81
          GenBank mRNA AK013037.1 - 1 - 2 620 81
          BC027683.1 - 1 - 2 623 1345974 81
          BQ550868.1 - 2 - 3 249 81
          DV654233.1 - 2 - 3 341 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004327.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp688

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25505557 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25505562 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25505560 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1971759 BC027683.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1971759 BC038680.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1971759 BC052512.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1971759 AI447502.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1971759 NM_026999.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2010966 AK013037.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 734831 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 76364 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 174977 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P131994 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2010966 AK013037.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1971759 BC027683.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1971759 BC038680.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1971759 BC052512.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1971759 AI447502.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1971759 NM_026999.4 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505563 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505576 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505566 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505556 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505544 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505550 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505570 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505562 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505543 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505579 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505575 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505572 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505551 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505547 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505557 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505581 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505569 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505571 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505553 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505583 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505546 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505573 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505554 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505568 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P131994 - -
          Whole Mouse Genome G4122A A_51_P355821 BC027683.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P355821 BC038680.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P355821 BC052512.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P355821 AI447502.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P355821 NM_026999.4 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121627 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P131994 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06031468 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06031465 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06031472 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06031469 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06031473 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06031466 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25505560 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25505562 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25505557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505546 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505559 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505562 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06212350 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505548 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505551 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505583 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505542 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06477404 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06638638 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505556 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06031474 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505567 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06352430 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06395895 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05988385 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505581 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06031473 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06477403 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505568 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505544 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505547 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505563 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505569 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06031467 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10568248 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568251 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568249 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568250 - -
          Affymetrix HT MG-430 PM Array Plate 1452588_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10568247 - -
          Mouse Genome 430 2.0 Array 1452588_PM_at - -
          Mouse Genome 430 2.0 Array 1452588_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69234_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69234_at - -
          Affymetrix Mouse Clariom S Array PSR0700036350.mm.1 NM_026999.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036350.mm.1 NM_026999.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036350.mm.1 TC0700004327.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700036349.mm.1 TC0700004327.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700036343.mm.1 NM_026999.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036343.mm.1 NM_026999.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036343.mm.1 TC0700004327.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700036346.mm.1 NM_026999.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036346.mm.1 NM_026999.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036346.mm.1 TC0700004327.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700036351.mm.1 NM_026999.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036351.mm.1 NM_026999.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036351.mm.1 TC0700004327.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700036348.mm.1 TC0700004327.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700036341.mm.1 NM_026999.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036341.mm.1 NM_026999.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036341.mm.1 TC0700004327.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700036342.mm.1 NM_026999.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036342.mm.1 NM_026999.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036342.mm.1 TC0700004327.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700036352.mm.1 NM_026999.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036352.mm.1 NM_026999.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036352.mm.1 TC0700004327.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700036347.mm.1 NM_026999.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036347.mm.1 NM_026999.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036347.mm.1 TC0700004327.mm.1 MTA TC
          Murine Genome U74v2 Set 103631_at - -
          Murine Genome U74v2 Set 103631_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1452588_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1452588_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 912960 - -
          ABI Mouse Genome Survey Microarray 932416 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101980019 - -
          Illumina mouse-6 v1.0 expression beadchip 101980019 - -
          Illumina mouse-6 v1.0 expression beadchip 510280 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000510280 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1244649 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1244649 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2474725 - -
          mouse-6 v1.1 (Illumina) scl30645.3.1_18-S - -
          mouse-6 v1.1 (Illumina) scl000266.1_16-S - -
          Illumina Sentrix mouse-6.v1 bead array 510280 - -
          Illumina Sentrix mouse-6.v1 bead array 101980019 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl30645.3.1_18-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 510280 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101980019 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1244649 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1244649 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline