Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00510020_m1
          See other LRRC46 GE Assays ›
          Gene Symbol:
          Lrrc46
          Gene Name:
          leucine rich repeat containing 46
          Gene Aliases:
          1700006D24Rik
          Chromosome Location:
          Chr.11: 97034602 - 97041369 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00510020_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lrrc46
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027026.2 NP_081302.2 3 - 4 502 72
          GenBank mRNA AK005668.1 - 3 - 4 478 72
          AK158517.1 - 3 - 4 549 72
          BC022723.1 - 3 - 4 502 4503387 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100003634.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lrrc46

          Panther Classification:

          Molecular Function -

          protein-binding activity modulator phosphatase modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of protein stability
          positive regulation of protein targeting to mitochondrion
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28048327 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28048328 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P375546 AK158517.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P375546 BC022723.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P375546 NM_027026.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P375546 AK005668.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P375546 AK158517.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P375546 BC022723.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P375546 NM_027026.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P375546 AK005668.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 467098 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 468400 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 551138 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 123522 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P166408 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P375546 AK158517.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P375546 BC022723.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P375546 NM_027026.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P375546 AK005668.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P375546 AK158517.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P375546 BC022723.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P375546 NM_027026.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P375546 AK005668.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28048293 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28048310 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28048312 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28048317 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28048328 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28048301 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28048297 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28048327 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28048316 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28048300 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28048318 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28048296 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28048325 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28048322 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28048299 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28048315 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28048314 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28048324 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P166408 - -
          Whole Mouse Genome G4122A A_52_P375546 AK158517.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P375546 BC022723.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P375546 NM_027026.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P375546 AK005668.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P375546 AK158517.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P375546 BC022723.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P375546 NM_027026.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P375546 AK005668.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P216085 AK158517.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P216085 BC022723.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P216085 NM_027026.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P216085 AK005668.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117182 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135962 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P166408 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09420564 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09420561 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09202623 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28048328 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28048327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28048271 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28048312 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28048286 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28048290 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28048314 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09344523 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28048292 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28048316 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28048280 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28048308 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09202623 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28048300 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09653034 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09653017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28048310 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28048325 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28048297 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09344519 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28048281 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28048320 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28048293 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28048319 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09491379 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28048287 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28048327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28048301 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09653028 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28048288 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10390325 - -
          Affymetrix Mouse Gene 1.0 ST Array 10390326 - -
          Affymetrix Mouse Gene 1.0 ST Array 10390321 - -
          Affymetrix Mouse Gene 1.0 ST Array 10390320 - -
          Affymetrix Mouse Gene 1.0 ST Array 10390322 - -
          Affymetrix Mouse Gene 1.0 ST Array 10390324 - -
          Affymetrix Mouse Gene 1.0 ST Array 10390327 - -
          Affymetrix Mouse Gene 1.0 ST Array 10390323 - -
          Affymetrix HT MG-430 PM Array Plate 1424909_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10390319 - -
          Mouse Genome 430 2.0 Array 1424909_at - -
          Mouse Genome 430 2.0 Array 1424909_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69297_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69297_at - -
          Affymetrix Mouse Clariom S Array PSR1100033332.mm.1 NM_027026.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100033332.mm.1 NM_027026.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100033332.mm.1 TC1100003634.mm.1 MTA TC
          Mouse Expression Set 430 1424909_PM_at - -
          Mouse Expression Set 430 1424909_at - -
          Mouse Genome 430A 2.0 Array 1424909_at - -
          Mouse Genome 430A 2.0 Array 1424909_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424909_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424909_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 452218 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4850433 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004850433 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2737412 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2737412 - -
          mouse-6 v1.1 (Illumina) scl39656.9.1_122-S - -
          Illumina Sentrix mouse-6.v1 bead array 4850433 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl39656.9.1_122-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4850433 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2737412 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2737412 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0