Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00510116_s1
          See other WDR5B GE Assays ›
          Gene Symbol:
          Wdr5b
          Gene Name:
          WD repeat domain 5B
          Gene Aliases:
          2310009C03Rik, AI606931
          Chromosome Location:
          Chr.16: 36041190 - 36042974 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00510116_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Wdr5b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027113.2 NP_081389.1 1 836 87
          GenBank mRNA AK009247.1 - 1 751 87
          AK149554.1 - 807 87
          AK165100.1 - 700 87
          BC064045.1 - 1 836 1245342 87
          EF011616.1 - 1 834 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600000502.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Wdr5b

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30673432 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30673428 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30673431 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30673429 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30673430 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P281711 BC064045.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P281711 EF011616.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P281711 AK149554.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P281711 AK165100.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P281711 AK009247.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P281711 BC064045.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P281711 EF011616.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P281711 AK149554.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P281711 AK165100.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P281711 AK009247.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 421325 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 937903 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P141628 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P129596 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P173204 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P281711 BC064045.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P281711 EF011616.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P281711 AK149554.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P281711 AK165100.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P281711 AK009247.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P281711 BC064045.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P281711 EF011616.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P281711 AK149554.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P281711 AK165100.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P281711 AK009247.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673394 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673436 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673414 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673432 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673423 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673406 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673411 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673437 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673445 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673405 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673424 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673422 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673407 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673391 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673426 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673397 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673408 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673442 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673429 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673403 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673415 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673444 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673446 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673416 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673393 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673419 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673440 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673400 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673410 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673447 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673427 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673434 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673418 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673392 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P173204 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P129596 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P141628 - -
          Whole Mouse Genome G4122A A_52_P281711 BC064045.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P281711 EF011616.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P281711 AK149554.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P281711 AK165100.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P281711 AK009247.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P281711 BC064045.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P281711 EF011616.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P281711 AK149554.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P281711 AK165100.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P281711 AK009247.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103506 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P141628 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P173204 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P129596 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12836309 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12624053 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12624073 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12836308 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12624079 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30673431 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30673432 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30673428 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30673429 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30673430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673427 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673436 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673391 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673397 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673444 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12872511 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673442 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673426 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12872510 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673419 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673393 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673423 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673420 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673434 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673445 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673416 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673437 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12624083 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673408 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12624079 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673406 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673429 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673407 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673427 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673422 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673431 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673415 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673410 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673392 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12895430 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673446 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12624061 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673424 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673405 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673414 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673400 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673447 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12685388 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10435488 - -
          Affymetrix HT MG-430 PM Array Plate 1453058_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10435487 - -
          Mouse Genome 430 2.0 Array 1453058_PM_at - -
          Mouse Genome 430 2.0 Array 1453058_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69544_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69544_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1600004819.mm.1 NM_027113.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600004819.mm.1 NM_027113.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600004819.mm.1 TC1600000502.mm.1 MTA TC
          Mouse Expression Set 430 1453058_PM_at - -
          Mouse Expression Set 430 1453058_at - -
          Affymetrix HT Mouse Genome 430B Array 1453058_at - -
          Affymetrix HT Mouse Genome 430B Array 1453058_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 571305 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 670110 - -
          Illumina mouse-6 v1.0 expression beadchip 670110 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000670110 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2425488 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2425488 - -
          mouse-6 v1.1 (Illumina) scl49184.1.1_144-S - -
          Illumina Sentrix mouse-6.v1 bead array 670110 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl49184.1.1_144-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 670110 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2425488 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2425488 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline