Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00510158_m1
          See other AP5S1 GE Assays ›
          Gene Symbol:
          Ap5s1
          Gene Name:
          adaptor-related protein 5 complex, sigma 1 subunit
          Gene Aliases:
          0610038L13Rik, 2310035K24Rik, AW060727, sigma5
          Chromosome Location:
          Chr.2: 131210360 - 131213514 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00510158_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ap5s1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001291031.1 NP_001277960.1 2 - 3 531 68
          NM_001291032.1 NP_001277961.1 2 - 3 531 68
          NM_027129.3 NP_081405.2 2 - 3 531 68
          GenBank mRNA AK002807.1 - 2 - 3 456 68
          AK009634.1 - 2 - 3 468 68
          BC048399.1 - 2 - 3 451 6438951 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200002011.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ap5s1

          Gene Ontology Categories:

          Function(s) Process(es)

          double-strand break repair via homologous recombination
          DNA repair
          transport
          cellular response to DNA damage stimulus
          protein transport
          endosomal transport
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21633884 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21633886 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21633885 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21633883 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2002319 BC048399.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2002319 NM_027129.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 668377 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 400806 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2002319 BC048399.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2002319 NM_027129.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21633899 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21633893 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21633904 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21633889 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21633895 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21633861 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21633902 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21633848 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21633872 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21633854 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21633860 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21633880 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21633881 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21633865 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21633871 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21633878 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21633852 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21633856 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21633858 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21633894 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21633859 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21633891 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21633884 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21633867 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21633874 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21633863 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21633869 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21633849 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21633876 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21633862 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21633887 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21633886 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04694200 - -
          Whole Mouse Genome G4122A A_51_P454002 BC048399.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P454002 AK002807.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P454002 NM_027129.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125448 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01411054 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01411040 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01411049 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01411055 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01411052 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21633883 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21633886 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21633885 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21633884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633831 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01411047 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633848 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01929102 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02041043 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633939 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633865 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01630934 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633936 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633933 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633836 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633878 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01411049 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01596614 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633840 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02282712 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633907 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01929105 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633894 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02026164 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01596619 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633924 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633869 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633922 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02282713 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02280595 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02280594 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01222908 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02282710 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633916 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633900 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01411052 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01845706 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633867 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01222904 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633861 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633891 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633932 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02041042 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633889 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01222914 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633863 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01601374 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633838 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633837 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633862 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01411040 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633909 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633904 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633912 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02098841 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01411054 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01222913 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633919 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01601373 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02041041 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633915 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633942 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633893 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01310662 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01411048 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633875 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633910 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01222918 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02282711 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10476271 - -
          Affymetrix Mouse Gene 1.0 ST Array 10476272 - -
          Affymetrix Mouse Gene 1.0 ST Array 10476275 - -
          Affymetrix Mouse Gene 1.0 ST Array 10476273 - -
          Affymetrix HT MG-430 PM Array Plate 1428241_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1432295_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10476270 - -
          Mouse Genome 430 2.0 Array 1428241_PM_at - -
          Mouse Genome 430 2.0 Array 1432295_a_at - -
          Mouse Genome 430 2.0 Array 1432295_PM_a_at - -
          Mouse Genome 430 2.0 Array 1428241_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69596_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69596_at - -
          Affymetrix Mouse Clariom S Array PSR0200015976.mm.1 NM_027129.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200015976.mm.1 TC0200002011.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200015977.mm.1 NM_027129.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200015977.mm.1 TC0200002011.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200015975.mm.1 NM_027129.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200015975.mm.1 TC0200002011.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200015984.mm.1 NM_027129.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200015984.mm.1 TC0200002011.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200015985.mm.1 NM_027129.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200015985.mm.1 TC0200002011.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200015979.mm.1 NM_027129.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200015979.mm.1 TC0200002011.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200015980.mm.1 NM_027129.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200015980.mm.1 TC0200002011.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200015982.mm.1 NM_027129.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200015982.mm.1 TC0200002011.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200015983.mm.1 NM_027129.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200015983.mm.1 TC0200002011.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200015974.mm.1 NM_027129.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200015974.mm.1 TC0200002011.mm.1 MTA TC
          Mouse Expression Set 430 1432295_a_at - -
          Mouse Expression Set 430 1432295_PM_a_at - -
          Murine Genome U74v2 Set 96819_PM_at - -
          Murine Genome U74v2 Set 96819_at - -
          Murine Genome U74v2 Set 166179_PM_f_at - -
          Murine Genome U74v2 Set 166179_f_at - -
          Mouse Genome 430A 2.0 Array 1432295_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1432295_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1432295_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1428241_at - -
          Affymetrix HT Mouse Genome 430A Array 1432295_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1428241_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 336971 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5360358 - -
          Illumina mouse-6 v1.0 expression beadchip 5360358 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005360358 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2646771 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2646771 - -
          mouse-6 v1.1 (Illumina) scl20267.4_3-S - -
          Illumina Sentrix mouse-6.v1 bead array 5360358 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl20267.4_3-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5360358 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2646771 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2646771 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl20267.4_3-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline