Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00510186_g1
          See other MIF4GD GE Assays ›
          Gene Symbol:
          Mif4gd
          Gene Name:
          MIF4G domain containing
          Gene Aliases:
          1110014L05Rik, 2310075G12Rik
          Chromosome Location:
          Chr.11: 115607918 - 115612973 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00510186_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mif4gd
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001243584.1 NP_001230513.1 3 - 4 428 61
          NM_001243586.1 NP_001230515.1 2 - 3 712 61
          NM_001243587.1 NP_001230516.1 3 - 4 428 61
          NM_027162.4 NP_081438.4 2 - 3 712 61
          GenBank mRNA AK003708.1 - 489 61
          AK010175.1 - 3 - 4 382 61
          AK150749.1 - 3 - 4 382 61
          BC019515.1 - 3 - 4 339 4460171 61
          BC026740.1 - 3 - 4 388 4015105 61
          BC026745.1 - 3 - 4 346 61
          BC055812.1 - 2 - 3 712 4934728 61
          CX210298.1 - 3 - 4 412 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100004299.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK003708.1,BC019515.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mif4gd

          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of translation
          RNA binding
          protein C-terminus binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100038382.mm.1 NM_001243584.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100038382.mm.1 NM_001243584.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100038382.mm.1 NM_001243586.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100038382.mm.1 NM_001243586.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100038382.mm.1 NM_001243587.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100038382.mm.1 NM_001243587.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100038382.mm.1 NM_027162.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100038382.mm.1 NM_027162.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100038382.mm.1 TC1100004299.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100038383.mm.1 NM_001243584.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100038383.mm.1 NM_001243584.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100038383.mm.1 NM_001243586.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100038383.mm.1 NM_001243586.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100038383.mm.1 NM_001243587.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100038383.mm.1 NM_001243587.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100038383.mm.1 NM_027162.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100038383.mm.1 NM_027162.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100038383.mm.1 TC1100004299.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline