Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00510275_m1
          See other APITD1 GE Assays ›
          Gene Symbol:
          Apitd1
          Gene Name:
          apoptosis-inducing, TAF9-like domain 1
          Gene Aliases:
          2610040C18Rik, 2810407L01Rik, CENP-S
          Chromosome Location:
          Chr.4: 149128349 - 149137600 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00510275_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Apitd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027263.2 NP_081539.1 1 - 2 123 60
          GenBank mRNA AK011725.1 - 1 - 2 72 60
          BC048521.1 - 1 - 2 100 6741262 60
          BC138926.1 - 1 - 2 64 8861948 60
          BC138927.1 - 1 - 2 64 8861949 60
          DV656255.1 - 1 - 2 123 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400004032.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Apitd1

          Panther Classification:

          Molecular Function -

          neuropeptide

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA repair
          cellular response to DNA damage stimulus
          cell cycle
          mitotic nuclear division
          cell division
          DNA binding
          chromatin binding
          double-stranded DNA binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23398854 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23398857 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23398855 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23398856 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P270519 AK012995.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P270519 AK013038.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P270519 NM_027263.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P270519 AK011725.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P270519 BC138926.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P270519 AK012995.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P270519 AK013038.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P270519 NM_027263.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P270519 AK011725.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P270519 BC138926.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P206058 BC048521.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P206058 BC048521.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 278899 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 164285 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 959881 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 472059 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 770550 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 168195 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P162883 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P270519 AK012995.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P270519 AK013038.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P270519 NM_027263.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P270519 AK011725.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P270519 BC138926.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P270519 AK012995.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P270519 AK013038.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P270519 NM_027263.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P270519 AK011725.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P270519 BC138926.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23398818 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23398815 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23398853 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23398863 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23398862 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23398807 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23398822 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23398814 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23398855 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23398828 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23398859 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23398833 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23398857 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23398831 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23398826 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23398864 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23398860 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23398821 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23398834 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23398825 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23398817 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23398850 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23398812 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23398811 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23398810 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23398808 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01015168 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P162883 - -
          Whole Mouse Genome G4122A A_51_P270519 AK012995.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P270519 AK013038.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P270519 NM_027263.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P270519 AK011725.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P270519 BC138926.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P270519 AK012995.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P270519 AK013038.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P270519 NM_027263.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P270519 AK011725.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P270519 BC138926.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P206058 BC048521.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P206058 BC048521.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104536 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P162883 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03829611 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03959976 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23398857 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23398854 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23398856 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23398855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398876 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398810 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03433514 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03817027 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398845 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398847 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398864 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398885 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398857 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03317258 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398825 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03946836 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398811 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398839 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398812 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398833 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398831 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03959975 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398876 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398815 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398821 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398848 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398840 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398879 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398880 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04008575 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398822 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398877 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398826 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398843 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03203852 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398861 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03317902 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398858 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398841 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03817024 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398844 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398832 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398835 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03358823 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398808 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398862 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398828 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398850 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398836 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398838 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03663397 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398855 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10518567 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518566 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518563 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518565 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518562 - -
          Affymetrix HT MG-430 PM Array Plate 1453067_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10518561 - -
          Mouse Genome 430 2.0 Array 1453067_at - -
          Mouse Genome 430 2.0 Array 1453067_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69928_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69928_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400033396.mm.1 TC0400004032.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400033396.mm.1 NM_027263.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400033396.mm.1 NM_027263.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400033390.mm.1 TC0400004032.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400033393.mm.1 TC0400004032.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400033393.mm.1 NM_027263.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400033393.mm.1 NM_027263.2 Refseq
          Mouse Expression Set 430 1453067_at - -
          Mouse Expression Set 430 1453067_PM_at - -
          Murine Genome U74v2 Set 167777_r_at - -
          Murine Genome U74v2 Set 167777_PM_r_at - -
          Affymetrix HT Mouse Genome 430B Array 1453067_at - -
          Affymetrix HT Mouse Genome 430B Array 1453067_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 688988 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2260458 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002260458 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1231416 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1231416 - -
          mouse-6 v1.1 (Illumina) scl23502.6.1_40-S - -
          Illumina Sentrix mouse-6.v1 bead array 2260458 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl23502.6.1_40-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2260458 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2983714 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2983714 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline