Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00510308_m1
          See other METTL18 GE Assays ›
          Gene Symbol:
          Mettl18
          Gene Name:
          methyltransferase like 18
          Gene Aliases:
          2810422O20Rik, C78571
          Chromosome Location:
          Chr.1: 163994891 - 163997243 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00510308_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mettl18
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK013137.1 - 1 - 2 67 73
          BC038687.1 - 1 - 2 40 73

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK013137.1 transcripts.

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20786428 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20786429 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20786430 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1953157 BC050143.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1953157 BC052693.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1953157 NM_027279.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1953157 AK013137.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1953157 BC038687.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2179726 BC050143.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2179726 BC052693.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2179726 NM_027279.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2179726 AK013137.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2179726 BC038687.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 751545 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 25908 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P155621 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2179726 BC050143.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2179726 BC052693.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2179726 NM_027279.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2179726 AK013137.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2179726 BC038687.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1953157 BC050143.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1953157 BC052693.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1953157 NM_027279.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1953157 AK013137.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1953157 BC038687.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20786452 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20786440 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20786436 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20786431 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20786430 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20786444 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20786445 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20786465 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20786453 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20786446 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20786428 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20786450 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20786457 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20786456 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20786443 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20786459 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20786433 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20786434 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20786460 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20786448 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20786466 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P155621 - -
          Whole Mouse Genome G4122A A_51_P442838 BC050143.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P442838 BC052693.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P442838 NM_027279.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P442838 AK013137.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P442838 AK139786.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126068 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P155621 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00063723 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00225624 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00225616 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00225621 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20786428 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20786429 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20786430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20786494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20786436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20786456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20786503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20786492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20786458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20786444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20786507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20786427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20786505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20786438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20786497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20786476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20786457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20786481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20786496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20786485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20786469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20786508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20786478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20786450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20786452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20786468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20786484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20786486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20786499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20786488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20786495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20786491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20786443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20786487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20786434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20786465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20786433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20786448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20786445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20786466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20786460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20786428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20786430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20786509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20786440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20786473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20786431 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00222877 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00225618 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20786457 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20786431 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20786434 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20786469 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00974169 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20786446 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20786465 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20786456 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20786450 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00551214 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20786445 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20786444 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20786466 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00730466 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20786460 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20786468 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20786480 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20786448 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20786430 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20786428 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20786476 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20786463 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00736853 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00063724 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00222878 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20786440 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20786470 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20786458 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20786436 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00974166 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10351181 - -
          Affymetrix Mouse Gene 1.0 ST Array 10351180 - -
          Affymetrix HT MG-430 PM Array Plate 1429137_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10351179 - -
          Mouse Genome 430 2.0 Array 1429137_at - -
          Mouse Genome 430 2.0 Array 1429137_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69962_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69962_PM_at - -
          Mouse Expression Set 430 1429137_PM_at - -
          Mouse Expression Set 430 1429137_at - -
          Murine Genome U74v2 Set 160999_r_at - -
          Murine Genome U74v2 Set 160999_PM_r_at - -
          Mouse Genome 430A 2.0 Array 1429137_PM_at - -
          Mouse Genome 430A 2.0 Array 1429137_at - -
          Affymetrix HT Mouse Genome 430A Array 1429137_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1429137_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 711539 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6650377 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006650377 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2625188 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2625188 - -
          mouse-6 v1.1 (Illumina) scl17289.2_452-S - -
          Illumina Sentrix mouse-6.v1 bead array 6650377 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl17289.2_452-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6650377 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2625188 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2625188 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline