Search
Search

Dclk2| Gene Symbol: Dclk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195496.1 | NP_001182425.1 | 9 - 10 | 2058 | 97 | |
| NM_001195497.1 | NP_001182426.1 | 8 - 9 | 2010 | 97 | ||
| NM_001195498.1 | NP_001182427.1 | 9 - 10 | 2061 | 97 | ||
| NM_001195499.1 | NP_001182428.1 | 9 - 10 | 2058 | 97 | ||
| NM_001195500.1 | NP_001182429.1 | 8 - 9 | 2010 | 97 | ||
| NM_027539.5 | NP_081815.3 | 8 - 9 | 2013 | 97 | ||
| GenBank mRNA | AB198721.1 | - | 9 - 10 | 1415 | 97 | |
| AK049126.1 | - | 8 - 9 | 1500 | 97 | ||
| AK049179.1 | - | 8 - 9 | 1462 | 97 | ||
| AK082633.1 | - | 8 - 9 | 1509 | 97 | ||
| AY968049.1 | - | 9 - 10 | 1346 | 97 | ||
| AY968050.1 | - | 9 - 10 | 1349 | 97 | ||
| AY968051.1 | - | 9 - 10 | 1346 | 97 | ||
| BC056921.1 | - | 8 - 9 | 1981 | 6822681 | 97 | |
| BC060192.1 | - | 8 - 9 | 1314 | 97 | ||
Set Membership: |
|