Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00510671_m1
          See other 4931428L18RIK GE Assays ›
          Gene Symbol:
          4931428L18Rik
          Gene Name:
          RIKEN cDNA 4931428L18 gene
          Gene Aliases:
          4930563O14Rik
          Chromosome Location:
          Chr.1: 31141078 - 31222656 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00510671_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4931428L18Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033445.1 1 - 2 181 83
          GenBank mRNA AK016212.1 - 1 - 2 182 83
          AK016483.1 - 1 - 2 137 83
          AK162175.1 - 1 - 2 136 81
          BC147514.1 - 1 - 2 153 9056506 83
          BC147546.1 - 1 - 2 153 9056537 83
          BC172083.1 - 1 - 2 153 9054369 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100002127.mm.1

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Whole Mouse Genome G4122A A_52_P169260 BC128067.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P169260 AF545851.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P169260 BC117745.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P169260 BC117746.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P169260 NM_153601.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P397078 AK016483.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P397078 NR_033445.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P397078 AK016483.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P397078 NR_033445.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10353724 - -
          Affymetrix Mouse Gene 1.0 ST Array 10353725 - -
          Affymetrix HT MG-430 PM Array Plate 1432347_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1430898_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1430897_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10353721 - -
          Mouse Genome 430 2.0 Array 1430898_s_at - -
          Mouse Genome 430 2.0 Array 1432347_PM_at - -
          Mouse Genome 430 2.0 Array 1430897_PM_at - -
          Mouse Genome 430 2.0 Array 1430897_at - -
          Mouse Genome 430 2.0 Array 1432347_at - -
          Mouse Genome 430 2.0 Array 1430898_PM_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70988_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70988_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0100017123.mm.1 NR_033445.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100017123.mm.1 NR_033445.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100017123.mm.1 TC0100002127.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100017124.mm.1 NR_033445.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100017124.mm.1 NR_033445.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100017124.mm.1 TC0100002127.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100017126.mm.1 NR_033445.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100017126.mm.1 NR_033445.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100017126.mm.1 TC0100002127.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100017120.mm.1 NR_033445.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100017120.mm.1 NR_033445.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100017120.mm.1 TC0100002127.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100017119.mm.1 NR_033445.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100017119.mm.1 NR_033445.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100017119.mm.1 TC0100002127.mm.1 MTA TC
          Mouse Expression Set 430 1430897_PM_at - -
          Mouse Expression Set 430 1432347_at - -
          Mouse Expression Set 430 1430897_at - -
          Mouse Expression Set 430 1432347_PM_at - -
          Mouse Expression Set 430 1430898_PM_s_at - -
          Mouse Expression Set 430 1430898_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1430898_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1430897_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430897_at - -
          Affymetrix HT Mouse Genome 430B Array 1432347_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432347_at - -
          Affymetrix HT Mouse Genome 430B Array 1430898_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 791137 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2470500 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1251021 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1251021 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1251021 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1251021 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline