| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1800008675.mm.1 |
NM_027733.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800008675.mm.1 |
NM_027733.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800008675.mm.1 |
NM_029485.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800008675.mm.1 |
NM_029485.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800008675.mm.1 |
TC1800001178.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800008680.mm.1 |
NM_027733.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800008680.mm.1 |
NM_027733.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800008680.mm.1 |
NM_029485.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800008680.mm.1 |
NM_029485.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800008680.mm.1 |
TC1800001178.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800008678.mm.1 |
NM_027733.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800008678.mm.1 |
NM_027733.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800008678.mm.1 |
NM_029485.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800008678.mm.1 |
NM_029485.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800008678.mm.1 |
TC1800001178.mm.1 |
MTA TC |