| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10504421 |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1452993_PM_at |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1457660_PM_x_at |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10504419 |
- |
- |
| Mouse Genome 430 2.0 Array |
1452993_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1452993_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1457660_x_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1457660_PM_x_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
71406_PM_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
71406_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0400003174.mm.1 |
NR_033355.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400003174.mm.1 |
NR_033355.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400003174.mm.1 |
TC0400000451.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1457660_x_at |
- |
- |
| Mouse Expression Set 430 |
1452993_at |
- |
- |
| Mouse Expression Set 430 |
1457660_PM_x_at |
- |
- |
| Mouse Expression Set 430 |
1452993_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1452993_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1452993_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1457660_PM_x_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1457660_x_at |
- |
- |