Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00510771_m1
          See other GGT6 GE Assays ›
          Gene Symbol:
          Ggt6
          Gene Name:
          gamma-glutamyltransferase 6
          Gene Aliases:
          9030405D14Rik, AI427679, AW490214
          Chromosome Location:
          Chr.11: 72435243 - 72438566 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00510771_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ggt6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027819.2 NP_082095.2 1 - 2 241 67
          GenBank mRNA AK018485.1 - 1 - 2 243 67
          BC058747.1 - 1 - 2 241 6394797 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100000994.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ggt6

          Panther Classification:

          Molecular Function -

          hydrolase

          Gene Ontology Categories:

          Function(s) Process(es)

          glutathione metabolic process
          glutathione biosynthetic process
          biological_process
          molecular_function
          gamma-glutamyltransferase activity
          transferase activity
          transferase activity, transferring acyl groups
          hydrolase activity
          glutathione hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P440238 BC058747.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P440238 NM_027819.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P440238 AK018485.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P440238 BC055837.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P440238 BC058747.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P440238 NM_027819.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P440238 AK018485.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P440238 BC055837.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2058073 BC058747.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2058073 NM_027819.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2058073 BC055837.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 71782 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 549622 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 218428 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P101038 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2058073 BC058747.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2058073 NM_027819.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2058073 BC055837.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P440238 BC058747.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P440238 NM_027819.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P440238 AK018485.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P440238 BC055837.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P440238 BC058747.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P440238 NM_027819.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P440238 AK018485.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P440238 BC055837.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27911454 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27911427 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27911448 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27911458 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27911450 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27911440 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27911435 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27911445 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27911456 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27911436 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27911459 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27911451 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27911461 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27911443 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27911455 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27911441 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27911437 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27911444 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27911433 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27911460 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27911425 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27911439 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27911453 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27911431 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27911429 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P101038 - -
          Whole Mouse Genome G4122A A_51_P440238 BC058747.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P440238 NM_027819.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P440238 AK018485.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P440238 BC055837.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P440238 BC058747.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P440238 NM_027819.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P440238 AK018485.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P440238 BC055837.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105676 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P101038 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09435848 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09568058 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09435847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27911516 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09435849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09074994 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27911450 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27911430 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27911414 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27911448 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09240868 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27911431 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27911439 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27911433 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27911428 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27911435 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27911408 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09435846 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27911425 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27911421 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27911412 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27911424 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27911460 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27911429 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27911445 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27911443 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27911427 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27911455 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27911453 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27911456 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09572576 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27911406 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27911417 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27911459 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27911441 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27911451 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09298864 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10378085 - -
          Affymetrix Mouse Gene 1.0 ST Array 10378083 - -
          Affymetrix Mouse Gene 1.0 ST Array 10378084 - -
          Affymetrix Mouse Gene 1.0 ST Array 10378086 - -
          Affymetrix HT MG-430 PM Array Plate 1453587_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10378082 - -
          Mouse Genome 430 2.0 Array 1453587_PM_at - -
          Mouse Genome 430 2.0 Array 1453587_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71522_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71522_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100008873.mm.1 NM_027819.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008873.mm.1 NM_027819.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008873.mm.1 TC1100000994.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100008874.mm.1 NM_027819.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008874.mm.1 NM_027819.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008874.mm.1 TC1100000994.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100008872.mm.1 NM_027819.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008872.mm.1 NM_027819.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008872.mm.1 TC1100000994.mm.1 MTA TC
          Mouse Expression Set 430 1453587_at - -
          Mouse Expression Set 430 1453587_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453587_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453587_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 838717 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3120139 - -
          Illumina mouse-6 v1.0 expression beadchip 3120139 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003120139 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2610790 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2610790 - -
          mouse-6 v1.1 (Illumina) scl41312.4_596-S - -
          Illumina Sentrix mouse-6.v1 bead array 3120139 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl41312.4_596-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3120139 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2803258 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2610790 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2803258 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline