Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Susd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162913.1 | NP_001156385.1 | 8 - 9 | 1343 | 88 | |
NM_027890.5 | NP_082166.3 | 10 - 11 | 1703 | 88 | ||
GenBank mRNA | AK004703.2 | - | 10 - 11 | 1673 | 88 | |
AK028817.1 | - | 10 - 11 | 1679 | 88 | ||
AK033823.1 | - | 8 - 9 | 1316 | 88 | ||
AK037059.1 | - | 10 - 11 | 1685 | 88 | ||
AK037127.1 | - | 10 - 11 | 1654 | 88 | ||
AK038944.1 | - | 8 - 9 | 1971 | 88 | ||
AK042469.1 | - | 10 - 11 | 1687 | 88 | ||
AK128959.1 | - | 10 - 11 | 1675 | 88 | ||
AK167155.1 | - | 9 - 10 | 1885 | 88 | ||
BC026837.1 | - | 10 - 11 | 1656 | 4222578 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |