Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00510899_m1
          See other DPEP3 GE Assays ›
          Gene Symbol:
          Dpep3
          Gene Name:
          dipeptidase 3
          Gene Aliases:
          1700018F16Rik, AI507039, MBD-3, MLZ-136
          Chromosome Location:
          Chr.8: 105973520 - 105979419 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00510899_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dpep3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027960.2 NP_082236.1 7 - 8 1137 71
          GenBank mRNA AF488553.1 - 7 - 8 1018 71
          AK006085.1 - 7 - 8 1138 71
          BC051148.1 - 7 - 8 1147 6704315 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800002846.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dpep3

          Panther Classification:

          Molecular Function -

          protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          meiotic cell cycle
          protein binding
          peptidase activity
          metalloexopeptidase activity
          metallopeptidase activity
          dipeptidyl-peptidase activity
          hydrolase activity
          dipeptidase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26137132 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26137126 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26137129 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26137131 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26137128 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26137127 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P329818 AK006085.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P329818 BC051148.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P329818 NM_027960.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P329818 AF488553.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P329818 AK006085.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P329818 BC051148.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P329818 NM_027960.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P329818 AF488553.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 822826 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 488711 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 104705 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 440523 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 117335 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P118791 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P329818 AK006085.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P329818 BC051148.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P329818 NM_027960.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P329818 AF488553.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P329818 AK006085.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P329818 BC051148.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P329818 NM_027960.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P329818 AF488553.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P329818 AK006085.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P329818 BC051148.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P329818 NM_027960.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P329818 AF488553.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P329818 AK006085.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P329818 BC051148.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P329818 NM_027960.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P329818 AF488553.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137122 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137120 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137149 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137131 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137117 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137128 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137126 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137113 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137115 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137141 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137114 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137132 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137124 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137129 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137111 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137144 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137133 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137112 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137140 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137138 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137136 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137135 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P118791 - -
          Whole Mouse Genome G4122A A_51_P329818 AK006085.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P329818 BC051148.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P329818 NM_027960.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P329818 AF488553.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P329818 AK006085.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P329818 BC051148.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P329818 NM_027960.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P329818 AF488553.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108135 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P118791 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06818401 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06967747 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06719750 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06719739 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06719740 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06719744 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26137128 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26137127 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26137131 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26137126 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26137129 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26137132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137120 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137110 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137124 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137106 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07190638 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06967747 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137141 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137135 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07069492 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06719763 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137115 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137111 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137105 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137131 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07431488 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137099 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137144 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137122 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06950317 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137146 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137133 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137095 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06755988 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137117 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07019874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137126 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10581422 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581423 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581428 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581427 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581425 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581433 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581429 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581430 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581424 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581426 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581432 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581431 - -
          Affymetrix HT MG-430 PM Array Plate 1429035_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10581421 - -
          Mouse Genome 430 2.0 Array 1429035_PM_at - -
          Mouse Genome 430 2.0 Array 1429035_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71854_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71854_at - -
          Affymetrix Mouse Clariom S Array PSR0800021937.mm.1 NM_027960.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021937.mm.1 NM_027960.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021937.mm.1 TC0800002846.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800021936.mm.1 NM_027960.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021936.mm.1 NM_027960.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021936.mm.1 TC0800002846.mm.1 MTA TC
          Mouse Expression Set 430 1429035_at - -
          Mouse Expression Set 430 1429035_PM_at - -
          Murine Genome U74v2 Set 161474_PM_r_at - -
          Murine Genome U74v2 Set 161358_PM_r_at - -
          Murine Genome U74v2 Set 161474_r_at - -
          Murine Genome U74v2 Set 161358_r_at - -
          Murine Genome U74v2 Set 170160_PM_r_at - -
          Murine Genome U74v2 Set 170160_r_at - -
          Mouse Genome 430A 2.0 Array 1429035_at - -
          Mouse Genome 430A 2.0 Array 1429035_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1429035_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1429035_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 352654 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3710706 - -
          Illumina mouse-6 v1.0 expression beadchip 3710706 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003710706 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2685120 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2685120 - -
          mouse-6 v1.1 (Illumina) scl34382.10.1_100-S - -
          Illumina Sentrix mouse-6.v1 bead array 3710706 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl34382.10.1_100-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3710706 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2685120 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2685120 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline