Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00511029_m1
          See other NKIRAS2 GE Assays ›
          Gene Symbol:
          Nkiras2
          Gene Name:
          NFKB inhibitor interacting Ras-like protein 2
          Gene Aliases:
          2410003M04Rik, 4930527H08Rik, D630018G21Rik, KBRAS2
          Chromosome Location:
          Chr.11: 100622955 - 100627602 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00511029_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nkiras2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028024.2 NP_082300.1 3 - 4 409 71
          GenBank mRNA AK010371.1 - 3 - 4 409 71
          AK015918.1 - 414 71
          AK052645.1 - 3 - 4 409 71
          AK083475.1 - 3 - 4 452 71
          AK149696.1 - 3 - 4 553 71
          AK169878.1 - 3 - 4 434 71
          BC013469.1 - 2 - 3 362 4219892 71
          BC024398.1 - 2 - 3 352 5003001 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001629.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nkiras2

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          small GTPase mediated signal transduction
          nucleotide binding
          GTP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2158478 NM_177790.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2158478 AK083475.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2158478 AK010371.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2158478 NM_028024.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2158483 NM_177790.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2158483 BC013469.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2158483 BC024398.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2158483 AK083475.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2158483 AK149696.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 410770 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 200071 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 312489 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P108669 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158483 NM_177790.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158483 BC013469.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158483 BC024398.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158483 AK083475.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158483 AK149696.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158478 NM_177790.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158478 AK083475.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158478 AK010371.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158478 NM_028024.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28069115 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28069121 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28069130 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28069119 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28069124 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28069126 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28069128 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28069117 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28069129 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28069118 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28069116 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P108669 - -
          Whole Mouse Genome G4122A A_52_P145066 BC013469.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P145066 BC024398.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P145066 AK083475.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P145066 AK149696.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P145066 AK169878.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P500226 NM_177790.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P500226 BC013469.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P500226 BC024398.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P500226 AK083475.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P500226 AK149696.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140283 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P108669 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09451665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28069070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09628157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28069127 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28069118 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09542837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28069113 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28069134 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09518317 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28069112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28069140 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28069136 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09194803 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28069111 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28069128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28069139 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09451671 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28069109 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28069132 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28069145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28069115 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28069124 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09542838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28069142 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28069144 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09542843 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10381168 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381166 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381167 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381169 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381165 - -
          Affymetrix HT MG-430 PM Array Plate 1424416_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10381162 - -
          Mouse Genome 430 2.0 Array 1424416_at - -
          Mouse Genome 430 2.0 Array 1424416_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71966_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71966_at - -
          Affymetrix Mouse Clariom S Array PSR1100015108.mm.1 NM_028024.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015108.mm.1 NM_028024.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015108.mm.1 TC1100001629.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100015110.mm.1 NM_028024.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015110.mm.1 NM_028024.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015110.mm.1 TC1100001629.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100015109.mm.1 NM_028024.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015109.mm.1 NM_028024.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015109.mm.1 TC1100001629.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100015112.mm.1 NM_028024.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015112.mm.1 NM_028024.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015112.mm.1 TC1100001629.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100015111.mm.1 NM_028024.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015111.mm.1 NM_028024.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015111.mm.1 TC1100001629.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100015113.mm.1 NM_028024.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015113.mm.1 NM_028024.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015113.mm.1 TC1100001629.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100015114.mm.1 NM_028024.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015114.mm.1 NM_028024.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015114.mm.1 TC1100001629.mm.1 MTA TC
          Mouse Expression Set 430 1424416_PM_at - -
          Mouse Expression Set 430 1424416_at - -
          Murine Genome U74v2 Set 92584_PM_at - -
          Murine Genome U74v2 Set 92584_at - -
          Mouse Genome 430A 2.0 Array 1424416_at - -
          Mouse Genome 430A 2.0 Array 1424416_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424416_at - -
          Affymetrix HT Mouse Genome 430A Array 1424416_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 375837 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 580519 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000580519 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2623184 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2623184 - -
          mouse-6 v1.1 (Illumina) scl071966.3_0-S - -
          Illumina Sentrix mouse-6.v1 bead array 580519 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl071966.3_0-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 580519 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2623184 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2623184 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl071966.3_0-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline