Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00511132_m1
          See other FUNDC1 GE Assays ›
          Gene Symbol:
          Fundc1
          Gene Name:
          FUN14 domain containing 1
          Gene Aliases:
          1500005J14Rik, 1810033P05Rik
          Chromosome Location:
          Chr.X: 17519967 - 17572330 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00511132_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fundc1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028058.3 NP_082334.1 2 - 3 306 118
          GenBank mRNA AK005163.1 - 2 - 3 307 118
          AK007686.1 - 1 - 2 343 118
          AK168868.1 - 2 - 3 293 118
          BC012515.1 - 2 - 3 248 2651125 118
          BC030487.1 - 2 - 3 312 5374103 118

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00001900.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fundc1

          Gene Ontology Categories:

          Function(s) Process(es)

          mitophagy
          response to hypoxia
          autophagy
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P271725 AK005163.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P271725 AK005163.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 427527 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 226464 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 192276 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 615050 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 824160 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 803684 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 54091 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 842193 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 336676 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 146803 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 93080 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P126728 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P157878 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P271725 AK005163.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P271725 AK005163.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P271725 AK005163.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P271725 AK005163.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32280348 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32280370 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32280343 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32280363 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32280365 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32280372 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32280347 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32280352 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32280358 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32280355 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32280345 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32280359 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32280350 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32280368 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32280361 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32280367 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32280349 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32280342 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32280366 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32280344 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32280341 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P126728 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03647346 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03647355 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P157878 - -
          Whole Mouse Genome G4122A A_51_P271725 AK005163.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P271725 AK005163.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113426 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P157878 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P126728 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15214483 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15121890 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15121905 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15121897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32280379 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32280347 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32280340 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32280318 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32280334 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15121903 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15000297 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32280310 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32280372 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32280355 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32280326 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15121905 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32280344 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14530148 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32280320 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32280322 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32280306 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14771807 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32280323 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32280343 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32280370 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32280342 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32280345 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32280348 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32280350 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14956487 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32280341 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32280324 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32280308 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15121890 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32280363 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32280366 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32280367 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32280327 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32280328 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32280358 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32280314 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32280321 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15112137 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32280325 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32280317 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32280336 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32280338 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32280304 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32280349 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32280333 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32280332 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32280329 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15018527 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10603787 - -
          Affymetrix Mouse Gene 1.0 ST Array 10603788 - -
          Affymetrix Mouse Gene 1.0 ST Array 10603790 - -
          Affymetrix Mouse Gene 1.0 ST Array 10603786 - -
          Affymetrix Mouse Gene 1.0 ST Array 10603789 - -
          Affymetrix HT MG-430 PM Array Plate 1453369_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1424215_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10603785 - -
          Mouse Genome 430 2.0 Array 1453369_PM_a_at - -
          Mouse Genome 430 2.0 Array 1424215_PM_at - -
          Mouse Genome 430 2.0 Array 1424215_at - -
          Mouse Genome 430 2.0 Array 1453369_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72018_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72018_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0X00012128.mm.1 NM_028058.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00012128.mm.1 TC0X00001900.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00012130.mm.1 NM_028058.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00012130.mm.1 TC0X00001900.mm.1 MTA TC
          Mouse Expression Set 430 1453369_PM_a_at - -
          Mouse Expression Set 430 1453369_a_at - -
          Mouse Expression Set 430 1424215_PM_at - -
          Mouse Expression Set 430 1424215_at - -
          Murine Genome U74v2 Set 171287_PM_i_at - -
          Murine Genome U74v2 Set 171287_i_at - -
          Mouse Genome 430A 2.0 Array 1453369_a_at - -
          Mouse Genome 430A 2.0 Array 1424215_PM_at - -
          Mouse Genome 430A 2.0 Array 1453369_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1424215_at - -
          Affymetrix HT Mouse Genome 430A Array 1424215_at - -
          Affymetrix HT Mouse Genome 430A Array 1453369_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1453369_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1424215_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 905374 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6420647 - -
          Illumina mouse-6 v1.0 expression beadchip 6400332 - -
          Illumina mouse-6 v1.0 expression beadchip 6420647 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006400332 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006420647 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2762755 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1229913 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1229913 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2762755 - -
          mouse-6 v1.1 (Illumina) scl0002994.1_38-S - -
          mouse-6 v1.1 (Illumina) scl54373.7_168-S - -
          Illumina Sentrix mouse-6.v1 bead array 6400332 - -
          Illumina Sentrix mouse-6.v1 bead array 6420647 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl54373.7_168-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0002994.1_38-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6400332 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6420647 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1229913 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2958016 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2762755 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2958016 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2762755 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1229913 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl54373.7_168-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline