Search
Search

Igsf5| Gene Symbol: Igsf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177886.1 | NP_001171357.1 | 2 - 3 | 165 | 82 | |
| NM_001177887.1 | NP_001171358.1 | 2 - 3 | 169 | 82 | ||
| NM_028078.3 | NP_082354.1 | 2 - 3 | 162 | 82 | ||
| GenBank mRNA | AF537215.1 | - | 2 - 3 | 103 | 82 | |
| AK008060.1 | - | 2 - 3 | 133 | 82 | ||
| AK078934.1 | - | 2 - 3 | 133 | 82 | ||
| AK086973.1 | - | 2 - 3 | 166 | 82 | ||
| BC004806.1 | - | 2 - 3 | 115 | 3584645 | 82 | |
| BX511978.1 | - | 2 - 3 | 193 | 82 | ||
| BY033616.1 | - | 2 - 3 | 163 | 82 | ||
Set Membership: |
|