Search
Search

Tmem68| Gene Symbol: Tmem68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028097.3 | NP_082373.1 | 5 - 6 | 869 | 91 | |
| GenBank mRNA | AK008493.1 | - | 5 - 6 | 903 | 91 | |
| AK048210.1 | - | 5 - 6 | 869 | 91 | ||
| AK084881.1 | - | 5 - 6 | 903 | 91 | ||
| AK088600.1 | - | 6 - 7 | 1039 | 91 | ||
| AK147765.1 | - | 5 - 6 | 902 | 91 | ||
| AK162563.1 | - | 5 - 6 | 902 | 91 | ||
| AK165916.1 | - | 5 - 6 | 931 | 91 | ||
| AK166237.1 | - | 5 - 6 | 955 | 91 | ||
| BC016240.1 | - | 5 - 6 | 887 | 4506688 | 91 | |
Set Membership: |
|