Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00511726_s1
          See other 1700052K11RIK GE Assays ›
          Gene Symbol:
          1700052K11Rik
          Gene Name:
          RIKEN cDNA 1700052K11 gene
          Gene Aliases:
          AI840954
          Chromosome Location:
          Chr.11: 105179020 - 105181433 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00511726_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700052K11Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027956.1 1 58 80
          GenBank mRNA AK006771.1 - 1 59 80
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA NM_028539.1 - 59 Chr. - - [-] on Build 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100003906.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK006771.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 1700052K11Rik

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28094597 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28094594 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P134262 BC099431.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P134262 AK156756.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P134262 NR_027956.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P134262 AK006771.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P134262 BC099431.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P134262 AK156756.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P134262 NR_027956.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P134262 AK006771.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 127023 - -
          Whole Mouse Genome G4122A A_51_P134262 BC099431.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P134262 AK156756.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P134262 NR_027956.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P134262 AK006771.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P134262 BC099431.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P134262 AK156756.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P134262 NR_027956.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P134262 AK006771.1 mouse_transcriptome
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28094594 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28094597 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10381873 - -
          Affymetrix Mouse Gene 1.0 ST Array 10392038 - -
          Affymetrix Mouse Gene 1.0 ST Array 10392039 - -
          Affymetrix Mouse Gene 1.0 ST Array 10392036 - -
          Affymetrix Mouse Gene 1.0 ST Array 10392037 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381874 - -
          Affymetrix HT MG-430 PM Array Plate 1429136_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10392035 - -
          Mouse Genome 430 2.0 Array 1429136_PM_at - -
          Mouse Genome 430 2.0 Array 1429136_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73431_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73431_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100036480.mm.1 NR_027956.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100036480.mm.1 NR_027956.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100036480.mm.1 TC1100003906.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100036479.mm.1 NR_027956.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100036479.mm.1 NR_027956.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100036479.mm.1 TC1100003906.mm.1 MTA TC
          Mouse Expression Set 430 1429136_PM_at - -
          Mouse Expression Set 430 1429136_at - -
          Affymetrix HT Mouse Genome 430B Array 1429136_at - -
          Affymetrix HT Mouse Genome 430B Array 1429136_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 664681 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1740452 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline