Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00511766_g1
          See other THOC3 GE Assays ›
          Gene Symbol:
          Thoc3
          Gene Name:
          THO complex 3
          Gene Aliases:
          2410044K02Rik, AL033344
          Chromosome Location:
          Chr.13: 54458837 - 54468840 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00511766_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Thoc3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028597.3 NP_082873.2 4 - 5 822 114
          GenBank mRNA AK010668.1 - 4 - 5 802 114
          BC019603.1 - 4 - 5 796 2647864 114

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300002031.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Thoc3

          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA processing
          mRNA export from nucleus
          transport
          RNA splicing
          viral mRNA export from host cell nucleus
          mRNA transport
          RNA binding
          protein binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29049669 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29049671 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29049668 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29049670 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P869802 BC019603.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P869802 AK010668.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P869802 NM_028597.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P869802 BC019603.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P869802 AK010668.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P869802 NM_028597.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 187694 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 332228 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 450715 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 698935 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P157607 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P869802 BC019603.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P869802 AK010668.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P869802 NM_028597.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P869802 BC019603.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P869802 AK010668.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P869802 NM_028597.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29049659 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29049693 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29049683 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29049662 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29049677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29049671 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29049687 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29049689 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29049655 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29049684 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29049692 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29049682 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29049657 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29049670 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29049679 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29049664 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29049666 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29049674 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29049665 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29049660 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P157607 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07880241 - -
          Whole Mouse Genome G4122A A_52_P869802 BC019603.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P869802 AK010668.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P869802 NM_028597.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P869802 BC019603.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P869802 AK010668.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P869802 NM_028597.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124022 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P157607 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10560619 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10886978 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29049671 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29049669 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29049670 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29049668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29049573 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29049680 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29049678 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29049650 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29049703 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29049631 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29049674 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29049668 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10781253 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29049638 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29049671 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10781257 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10871175 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29049646 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29049659 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29049632 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29049686 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29049672 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29049700 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29049679 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10550031 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29049677 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29049644 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29049664 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29049630 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29049662 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29049682 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10845645 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29049636 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29049685 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10655970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29049652 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29049665 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10609793 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29049648 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29049675 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29049660 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29049643 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29049683 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10856953 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29049640 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29049635 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29049702 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29049692 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29049657 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29049689 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29049647 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29049696 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10409326 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409323 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409329 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409325 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409327 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409328 - -
          Affymetrix HT MG-430 PM Array Plate 1423970_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1423971_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10409322 - -
          Mouse Genome 430 2.0 Array 1423971_PM_at - -
          Mouse Genome 430 2.0 Array 1423971_at - -
          Mouse Genome 430 2.0 Array 1423970_at - -
          Mouse Genome 430 2.0 Array 1423970_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73666_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73666_at - -
          Affymetrix Mouse Clariom S Array PSR1300012656.mm.1 NM_028597.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300012656.mm.1 NM_028597.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300012656.mm.1 TC1300002031.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300012658.mm.1 NM_028597.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300012658.mm.1 NM_028597.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300012658.mm.1 TC1300002031.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300012659.mm.1 NM_028597.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300012659.mm.1 NM_028597.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300012659.mm.1 TC1300002031.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300012660.mm.1 NM_028597.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300012660.mm.1 NM_028597.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300012660.mm.1 TC1300002031.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300012665.mm.1 NM_028597.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300012665.mm.1 NM_028597.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300012665.mm.1 TC1300002031.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300012664.mm.1 NM_028597.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300012664.mm.1 NM_028597.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300012664.mm.1 TC1300002031.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300012663.mm.1 NM_028597.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300012663.mm.1 NM_028597.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300012663.mm.1 TC1300002031.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300012657.mm.1 NM_028597.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300012657.mm.1 NM_028597.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300012657.mm.1 TC1300002031.mm.1 MTA TC
          Mouse Expression Set 430 1423971_PM_at - -
          Mouse Expression Set 430 1423971_at - -
          Mouse Expression Set 430 1423970_at - -
          Mouse Expression Set 430 1423970_PM_at - -
          Murine Genome U74v2 Set 99490_PM_at - -
          Murine Genome U74v2 Set 99490_at - -
          Mouse Genome 430A 2.0 Array 1423971_PM_at - -
          Mouse Genome 430A 2.0 Array 1423970_PM_at - -
          Mouse Genome 430A 2.0 Array 1423971_at - -
          Mouse Genome 430A 2.0 Array 1423970_at - -
          Affymetrix HT Mouse Genome 430A Array 1423970_at - -
          Affymetrix HT Mouse Genome 430A Array 1423971_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423970_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423971_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6110142 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006110142 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2757197 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2757197 - -
          mouse-6 v1.1 (Illumina) scl43949.6.1_8-S - -
          Illumina Sentrix mouse-6.v1 bead array 6110142 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl43949.6.1_8-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6110142 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2888342 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2757197 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2757197 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2888342 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl43949.6.1_8-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline