Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00511786_m1
          See other GLIPR1 GE Assays ›
          Gene Symbol:
          Glipr1
          Gene Name:
          GLI pathogenesis-related 1 (glioma)
          Gene Aliases:
          2410114O14Rik, RTVP-1, RTVP1, mRTVP-1
          Chromosome Location:
          Chr.10: 111984792 - 112002619 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00511786_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Glipr1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028608.3 NP_082884.1 3 - 4 740 85
          GenBank mRNA AK010768.1 - 3 - 4 672 85
          AK137557.1 - 3 - 4 687 85
          AK170051.1 - 3 - 4 687 85
          BC025083.1 - 3 - 4 664 3711757 85
          BC098231.1 - 3 - 4 714 3325823 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000002878.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Glipr1

          Panther Classification:

          Molecular Function -

          defense/immunity protein

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P371051 AK170051.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P371051 BC025083.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P371051 BC098231.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P371051 NM_028608.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P371051 AK010768.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P371051 AK170051.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P371051 BC025083.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P371051 BC098231.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P371051 NM_028608.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P371051 AK010768.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 123054 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 114263 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 199789 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 736307 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 75497 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P100830 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P112071 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P371051 AK170051.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P371051 BC025083.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P371051 BC098231.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P371051 NM_028608.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P371051 AK010768.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P371051 AK170051.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P371051 BC025083.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P371051 BC098231.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P371051 NM_028608.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P371051 AK010768.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27452299 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27452297 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27452311 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27452318 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27452320 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27452306 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27452326 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27452324 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27452322 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27452314 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27452307 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27452309 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27452303 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27452313 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27452325 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27452301 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07194666 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07194653 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P112071 - -
          Whole Mouse Genome G4122A A_51_P371051 AK170051.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P371051 BC025083.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P371051 BC098231.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P371051 NM_028608.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P371051 AK010768.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P371051 AK170051.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P371051 BC025083.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P371051 BC098231.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P371051 NM_028608.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P371051 AK010768.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137027 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133411 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P112071 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08697426 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08634711 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08884419 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08884420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27452297 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08914951 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27452286 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27452267 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27452319 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27452332 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27452287 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27452314 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27452306 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27452270 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08914942 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08663660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27452317 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08663663 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27452311 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08774025 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27452309 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27452312 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27452297 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27452324 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27452284 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27452291 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27452326 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08680039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27452294 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27452289 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27452272 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27452330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27452337 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08795191 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27452307 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27452278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27452283 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27452338 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08482887 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27452275 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27452299 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27452325 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27452313 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27452290 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08680042 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27452276 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27452331 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08340143 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08207367 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08914949 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27452273 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10372413 - -
          Affymetrix Mouse Gene 1.0 ST Array 10372415 - -
          Affymetrix Mouse Gene 1.0 ST Array 10372412 - -
          Affymetrix Mouse Gene 1.0 ST Array 10372411 - -
          Affymetrix Mouse Gene 1.0 ST Array 10372414 - -
          Affymetrix Mouse Gene 1.0 ST Array 10372416 - -
          Affymetrix HT MG-430 PM Array Plate 1449746_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1424927_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10372410 - -
          Affymetrix Mouse Gene 1.1 ST Array 10366350 - -
          Mouse Genome 430 2.0 Array 1424927_at - -
          Mouse Genome 430 2.0 Array 1424927_PM_at - -
          Mouse Genome 430 2.0 Array 1449746_s_at - -
          Mouse Genome 430 2.0 Array 1449746_PM_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73690_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73690_at - -
          Affymetrix Mouse Clariom S Array PSR1000021736.mm.1 NM_028608.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000021736.mm.1 NM_028608.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000021736.mm.1 TC1000002878.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000021734.mm.1 NM_028608.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000021734.mm.1 NM_028608.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000021734.mm.1 TC1000002878.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000021739.mm.1 NM_028608.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000021739.mm.1 NM_028608.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000021739.mm.1 TC1000002878.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000021732.mm.1 NM_028608.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000021732.mm.1 NM_028608.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000021732.mm.1 TC1000002878.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000021735.mm.1 NM_028608.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000021735.mm.1 NM_028608.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000021735.mm.1 TC1000002878.mm.1 MTA TC
          Mouse Expression Set 430 1424927_PM_at - -
          Mouse Expression Set 430 1424927_at - -
          Mouse Expression Set 430 1449746_s_at - -
          Mouse Expression Set 430 1449746_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1424927_at - -
          Mouse Genome 430A 2.0 Array 1424927_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424927_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424927_at - -
          Affymetrix HT Mouse Genome 430B Array 1449746_PM_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1449746_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 673276 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3360750 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003360750 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1215807 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1215807 - -
          mouse-6 v1.1 (Illumina) scl37497.8.1_151-S - -
          Illumina Sentrix mouse-6.v1 bead array 3360750 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl37497.8.1_151-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3360750 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1215807 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline